Skip to Content
Merck
All Photos(1)

Key Documents

48953-U

Supelco

EPA 525 PAH Mix A

certified reference material, 500 μg/mL each component in dichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Agency

EPA 525

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

500 μg/mL each component in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

General description

EPA 525 PAH Mix A is a multi-component solution containing acenaphthylene, anthracene, benz[a]anthracene, benzo[b]fluoranthene, benzo[k]fluoranthene, benzo[ghi]perylene, benzo[a]pyrene, chrysene, dibenz[a,h]anthracene, fluorine, indeno[1,2,3-cd]pyrene, phenanthrene, and pyrene as certified reference materials (CRMs).
For use in extraction and quantification of general purpose organics.

Application

EPA 525 PAH Mix Standard A is used as a CRM to quantify polycyclic aromatic hydrocarbons (PAHs) in tree barks, particulate matter, gaseous samples, aerosols, and Porcellio scaber Latr. (Isopoda) by gas chromatography-mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acenaphthylene
    Anthracene
    Benz[a]anthracene
    Benzo[b]fluoranthene
    Benzo[k]fluoranthene
    Benzo[ghi]perylene
    Benzo[a]pyrene
    Chrysene
    Dibenz[a,h]anthracene
    Fluorene
    Indeno[1,2,3-cd]pyrene
    Phenanthrene
    Pyrene
    See All (13)

Signal Word

Danger

Hazard Statements

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polycyclic aromatic hydrocarbons in tree barks, gaseous and particulate phase samples collected near an industrial complex in Sao Paulo (Brazil)
Pereira GM, et al.
Chemosphere, 237(3), 124499-124499 (2019)
The effect of soil pollution with petroleum-derived substances on Porcellio scaber Latr.(Crustacea, Isopoda)
Gospodarek J, et al.
Environmental Monitoring and Assessment, 191(1), 38-38 (2019)
Biomass burning in the Amazon region: Aerosol source apportionment and associated health risk assessment
de Oliveira Alves N, et al.
Atmospheric Environment, 120(3), 277-285 (2015)
Organic compounds in particulate and gaseous phase collected in the neighbourhood of an industrial complex in S?o Paulo (Brazil)
Caumo S, et al.
Air quality, atmosphere, & health, 11(3), 271-283 (2018)
Milena Simões Peixoto et al.
Chemosphere, 231, 518-527 (2019-06-01)
Retene (RET) is the most abundant polycyclic aromatic hydrocarbon (PAH) released upon burning of cellulose, although it is not considered as one of the priority PAHs and is not included for risk assessments by the US Environmental Protection Agency (US-EPA).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service