Skip to Content
Merck
All Photos(1)

Key Documents

48498

Supelco

Bis-(2-chloroisopropyl) ether

analytical standard

Synonym(s):

2-Chloroisopropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12Cl2O
CAS Number:
Molecular Weight:
171.06
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

CC(CCl)OC(C)CCl

InChI

1S/C6H12Cl2O/c1-5(3-7)9-6(2)4-8/h5-6H,3-4H2,1-2H3

InChI key

QCFYJCYNJLBDRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis-(2-chloroisopropyl) ether is a colorless liquid, used as a solvent for greases, waxes and fats. It can also find applications in manufacturing of textiles, spotting agents and cleaning solutions.

Application

Bis-(2-chloroisopropyl) ether may be used as an analytical standard for the determination of the analyte in water by solid-phase microextraction and sewage sludge by gas chromatography/mass spectrometry (GC/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

185.0 °F

Flash Point(C)

85 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative determination of organic priority pollutants in sewage sludge by GC/MS
Lega R, et al.
Chemosphere, 34(8), 1705-1712 (1997)
Determination of haloethers in water by solid-phase microextraction
Huang SD, et al.
Journal of Chromatography A, 769(2), 239-246 (1997)
Saro Jayaraman et al.
Environmental science & technology, 43(21), 8387-8392 (2009-11-21)
While breeding tree swallows (Tachycineta bicolor) have been used as biomonitors for freshwater sites, we report the first use of this species to assess contaminant bioaccumulation from estuarine breeding grounds into these aerial insectivores. Eggs and nestlings were collected from
Orme-Zavaleta J and Connery J
Drinking Water Health Advisory: Volatile Organic Compounds (1990)
Bis(2-chloro-1-methylethyl)ether.
IARC monographs on the evaluation of carcinogenic risks to humans, 71 Pt 3, 1275-1279 (1999-09-07)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service