Skip to Content
Merck
All Photos(1)

Key Documents

27025

Supelco

Vials, clear glass, screw top, graduated (vial only)

volume 7 mL, thread for 15-425, pkg of 100 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
Pricing and availability is not currently available.

material

clear glass vial (with graduations)

description

cap not included

feature

closure type screw top vial
graduated

packaging

pkg of 100 ea

O.D. × H

17 mm × 60 mm

volume

7 mL

fitting

thread for 15-425

Looking for similar products? Visit Product Comparison Guide

General description

Vials are suitable basically to inject samples from an autosampler. Autosampler needle pierces through the cap during injection and withdraws the required aliquots of sample from the vials. Screw top vials (volume 7 mL, thread 15-425) are used on many autosamplers. Made from clear glass and includes graduations.

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Theresa J Barberi et al.
PloS one, 7(1), e29808-e29808 (2012-01-13)
Pin1 has previously been described to regulate cells that participate in both innate and adaptive immunity. Thus far, however, no role for Pin1 has been described in modulating conventional dendritic cells, innate antigen presenting cells that potently activate naïve T

Questions

  1. What is the tolerance for the markings and what are the dimensions of vials 27024, 27025, NS 27026?

    1 answer
    1. The vial markings serve as reference points, but they are not precise indicators of volume. The vial printing process is consistent; there is no 100% guarantee for accuracy. Below are the details for each vial:
      1. Vial no. 27025: Volume 7ml, OD (mm) 17, Height (mm) 60, Thread Size 15-425
      2. Vial no. 20724: Volume 4ml, OD (mm) 15, Height (mm) 45, Thread Size 13-425
      3. Vial no. 27026: Volume 15ml, OD (mm) 21, Height (mm) 70, Thread Size 18-400

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service