Skip to Content
Merck
All Photos(2)

Documents

WG128

Sigma-Aldrich

Wright-Giemsa Stain, Modified

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116121
NACRES:
NA.47

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

0.4 % (w/v) in methanol (buffered at pH 6.8)

pH

6.7-6.9

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

When blood films are stained using Wright-Giemsa Stain, the white blood cell nucleus and cytoplasm take on the characteristic blue or pink coloration. The combination of purified eosin and thiazine dyes in the product eliminates inconsistent staining and yields reproducible chromogenic responses.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Joseph M D Rastrick et al.
PloS one, 8(1), e54128-e54128 (2013-01-26)
COPD is an inflammatory lung disease largely associated with exposure to cigarette smoke (CS). The mechanism by which CS leads to the pathogenesis of COPD is currently unclear; it is known however that many of the inflammatory mediators present in
Barbora Bílková et al.
Veterinary immunology and immunopathology, 188, 71-77 (2017-06-16)
While haematological variation is well known in birds, variation in avian breeds (distinct morphotypes of the same species) remains unexplored. Poultry breeds, in particular, may show interesting evolutionary patterns and economically-relevant physiological differences. We performed a comparative examination of blood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service