Skip to Content
Merck
All Photos(5)

Documents

V5255

Sigma-Aldrich

Monoclonal Anti-Vimentin antibody produced in mouse

clone VIM-13.2, ascites fluid

Synonym(s):

Anti-F5H288

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

VIM-13.2, monoclonal

species reactivity

chicken, rat, human, rabbit, frog, mouse

technique(s)

indirect immunofluorescence: 1:200 using cultured chicken fibroblasts
western blot: 1:500 using human fibroblasts cell extract

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VIM(7431)
mouse ... Vim(22352)
rat ... Vim(81818)

General description

Vimentin is a 53 kD type III intermediate filaments expressed in breast myoepithelial cells, osteocytes, langerhans cells of the skin, schwann cells and astrocytes. Monoclonal anti-vimentin antibody can be used in immunofluorescent labeling of frozen tissue sections to localize vimentin in fibroblasts, endothelial cells, lymphoid tissues and melanocytes. It can also be used in western blotting. Mouse anti-vimentin antibody reacts specifically with vimentin. This product has also shown best reactivity with cultured chicken fibroblasts and can also react with human and mouse cells or tissues.The protein monomers consist of α-helical rod, flanked by head and tail domains. The VIM gene is mapped to human chromosome 10p13.

Immunogen

human foreskin fibroblasts

Application

Monoclonal anti-vimentin antibody can be used in the following:
  • Immunocytochemical localization of vimentin in normal and pathological tissue of mesenchymal origin.
  • It can also be used in immunoblotting
  • Immunofluorescence
  • Further, anti-vimentin antibody can be used to distinguish tumors and metastatic lesions derived from sarcomas, lymphomas, and melanomas.

Biochem/physiol Actions

Vimentin has a crucial role in maintaining cell shape and motility. It also provides mechanical stress tolerance for mesenchymal cells. Vimentin induces fibroblast motility and promotes wound healing process. Vimentin filaments form a part of cytoskeleton filament system and therefore, support and secure the cell organelles and also maintain cellular integrity. Vimentin is associated with the process of initiation, progression, metastasis and epithelial-mesenchymal transition of cancer cells, thereby contributing to tumorigenesis. Upregulation of this gene is observed in various cancers including prostate cancer, gastrointestinal tumors, lung cancer, breast cancer, CNS tumors and malignant melanoma.

Other Notes

This product can be found as purified product that was produced using cell culture hybridoma product.
SAB4200716 Anti-Vimentin antibody, Mouse monoclonal
clone VIM-13.2, purified from hybridoma cell culture

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lung cancer exosomes as drivers of epithelial mesenchymal transition.
Rahman M A, et al.
Oncotarget, 7(34), 54852-54852 (2016)
Yvonne Ting-Chun Yu et al.
Journal of biomedical science, 23, 14-14 (2016-01-24)
Severe acute respiratory syndrome coronavirus (SARS-CoV) caused a global panic due to its high morbidity and mortality during 2002 and 2003. Soon after the deadly disease outbreak, the angiotensin-converting enzyme 2 (ACE2) was identified as a functional cellular receptor in
A novel role of vimentin filaments; binding and stabilization of collagen mRNAs.
Challa A A and Stefanovic B
Molecular and Cellular Biology (2011)
Qing Wang et al.
BMC cancer, 13, 137-137 (2013-03-22)
We attempted to identify novel biomarkers and therapeutic targets for esophageal squamous cell carcinoma by gene expression profiling of frozen esophageal squamous carcinoma specimens and examined the functional relevance of a newly discovered marker gene, WDR66. Laser capture microdissection technique
Sang Goo Lee et al.
PloS one, 11(6), e0157768-e0157768 (2016-06-29)
Unlike mammals, the non-mammalian vertebrate inner ear can regenerate the sensory cells, hair cells, either spontaneously or through induction after hair cell loss, leading to hearing recovery. The mechanisms underlying the regeneration are poorly understood. By microarray analysis on a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service