Skip to Content
Merck
All Photos(1)

Key Documents

V3254

Supelco

Vitamin B12-Agarose

saline suspension

Synonym(s):

Cyanocobalamin-Agarose

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12190000
NACRES:
NA.21

form

saline suspension

extent of labeling

≥0.3 mg per mL

matrix

4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

carboxyl

matrix spacer

8 atoms

storage temp.

2-8°C

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam Schaffner et al.
Cell research, 29(4), 313-329 (2019-03-13)
Missense mutations in Leucine-Rich Repeat Kinase 2 (LRRK2) cause the majority of familial and some sporadic forms of Parkinson's disease (PD). The hyperactivity of LRRK2 kinase induced by the pathogenic mutations underlies neurotoxicity, promoting the development of LRRK2 kinase inhibitors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service