TRUE4
TrueGel3D Hydrogel Kits
FAST-PVA, CD cell-degradable crosslinker
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
shipped in
dry ice
storage temp.
−70°C
Looking for similar products? Visit Product Comparison Guide
General description
The gel is formed by stable thioether bonds between maleimide groups of PVA (FAST-PVA) and thiol groups on CD cell-degradable crosslinker. Malemide-functionalized PVA is specifically used to decrease the time for gel formation. The PVA polymer does not have cell attachment sites, but it can be customized to conjugate RGD peptide using TrueGel3D RGD peptide (TRUERGD-1EA) prior to the crosslinking step. The CD cell-degradable crosslinker is composed of matrix metalloprotease (MMP)-cleavable peptide (Pro-Leu-Gly-Leu-Trp-Ala), which allows cells to spread and migrate by secreting matrix metalloproteases (MMP1, MMP3, MMP7 and MMP9).
Application
TRUE4-1KT is used for cell migration studies of amoeboid cells including neutrophils, dendritic cells and lymphocytes. It has been preferred to use PVA-based gels over dextran-based gels for long-term culture of cells producing carbohydrate degrading enzymes.
Features and Benefits
- Fast gelling (takes few seconds to minutes after addition of crosslinker)
- Flexibility in modulating gel stiffness
- Contains MMP-cleavable crosslinker
- Cells cannot be recovered
- Cells do not attach to hydrogel (if not customized with TrueGel3D RGD peptide)
Components
TRUE4-1KT contains FAST-PVA polymer solution in phosphate buffer and lyophilized CD cell-degradable crosslinker with reconstitution buffer, TrueGel3D thioglycerol and water.
Caution
Avoid prolong exposure of CD cell-degradable crosslinker to air and store FAST-PVA in small portions to avoid frequent freeze-thaw cycles.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service