Skip to Content
Merck
All Photos(2)

Key Documents

T5534

Sigma-Aldrich

Glyceryl triundecanoate

≥98%

Synonym(s):

1,2,3-Triundecanoylglycerol, Glycerol triundecanoate, Triundecanoin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)9COOCH2]2CHOCO(CH2)9CH3
CAS Number:
Molecular Weight:
596.92
Beilstein:
1810343
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Quality Level

Assay

≥98%

form

solid

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCC)OC(=O)CCCCCCCCCC

InChI

1S/C36H68O6/c1-4-7-10-13-16-19-22-25-28-34(37)40-31-33(42-36(39)30-27-24-21-18-15-12-9-6-3)32-41-35(38)29-26-23-20-17-14-11-8-5-2/h33H,4-32H2,1-3H3

InChI key

MBXVIRZWSHICAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Glyceryl triundecanoate has been used:
  • as a reference standard in high-performance liquid chromatography (HPLC) analysis to identify the hydrolysis products and lipolytic activity in human milk
  • as an internal standard in gas chromatography analysis for total fat estimation
  • as an internal standard for fatty acid analysis in vegetable and meat samples

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jozef Kováčik et al.
Journal of plant physiology, 252, 153228-153228 (2020-07-24)
Responses of common medicinal herb Matricaria chamomilla to short-term cadmium (Cd) exposure (48 h) alone or in combination with nitric oxide (NO) scavenger (PTIO) or donor (SNP) were studied. Modulators revealed expected impact on NO formation (depletion under PTIO but
Nádia Costa et al.
Food control, 64, 128-134 (2016-06-09)
Consistent evidence exist on the harmful health effects of industrial trans fatty acids (TFA). In order to have accurate data on TFA intake and implement adequate measures to reduce their intake, each country should have updated estimates of TFA content
S Morera Pons et al.
Journal of chromatography. A, 823(1-2), 467-474 (1998-11-18)
The effect of various storage methods on the stability of the triacylglyceride fraction of human milk was evaluated. Samples were treated as follows: Group I--stored at -20 degrees C for 4 months, group II--heated for 1.5 min at 80 degrees
Jordi Eras et al.
Journal of chromatography. A, 1047(1), 157-161 (2004-10-16)
Acylglycerides present in oil seeds and meat can be transformed into volatile fatty esters using chlorotrimethylsilane (CTMS) and 1-pentanol as reagents. The volatile esters can then be analysed by GC. The method is quantitative and involves only minor sample manipulation.
Hongkai Xie et al.
Food & function, 11(3), 2349-2357 (2020-03-04)
The difference between the oxidative susceptibility of polyunsaturated fatty acids (PUFAs) esterified into triacylglycerols (TAG) and phospholipids (PL) and the effects of polyphenolic antioxidants on such susceptibility in dried seafood were investigated. Lipid fractions containing TAG and PL were extracted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service