Skip to Content
Merck
All Photos(1)

Key Documents

SRP6489

Sigma-Aldrich

Carbonic Anhydrase 4/CA4 human

recombinant, expressed in HEK 293 cells, ≥92% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.77

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥92% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 31 kDa
observed mol wt 33 kDa by SDS-PAGE (reducing) (Ala 19 is the predicted N-terminal .)

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CA4(762)

General description

Carbonic anhydrase 4 (CA4) is a 35kDa protein, encoded by the gene mapped to human chromosome 17q23.1-23.2. It is expressed on extracellular surfaces of certain endothelial and epithelial cells. CA4 is one among the 12 isozymes of carbonic anhydrase.
Carbonic anhydrase 4 (CA4) is also known as Carbonate dehydratase IV, Carbonic anhydrase IV, which belongs to the α-carbonic anhydrase family. CA4 may stimulate the sodium/bicarbonate transporter activity of SLC4A4 that acts in pH homeostasis. It is essential for acid overload removal from the retina and retina epithelium, and acid release in the choriocapillaris in the choroid. The enzyme regulation is activated by histamine, L-adrenaline, D-phenylalanine, L- and D-histidine. CA4 can interact with SLC4A4.

Biochem/physiol Actions

Carbonic anhydrase 4 (CA4) plays a vital role in CO2 hydration activity. It also catalyzes the dehydration of HCO3. In addition, it also facilitates CO2 and HCO3 transport in various vital tissues like kidney and lung Polymorphism in the gene has been associated with the development of sporadic retinitis pigmentosa (RP).

Physical form

Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7.5. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Membrane associated carbonic anhydrase IV (CA IV): a personal and historical perspective.
Waheed A, et al.
Sub-Cellular Biochemistry, 157-179 (2014)
Screening for the Carbonic Anhydrase IV Gene Mutations in Chinese Retinitis Pigmentosa Patients
Tian Y, et al.
Current Eye Research, 35(5), 440-444 (2010)
Li-Li Huang et al.
Journal of clinical and translational hepatology, 11(3), 605-613 (2023-03-28)
Hepatocellular carcinoma (HCC) is one of the most common types of cancer, often resulting in death. Augmenter of liver regeneration (ALR), a widely expressed multifunctional protein, has roles in liver disease. In our previous study, we reported that ALR knockdown
Familial microduplication of 17q23.1?q23.2 involving TBX4 is associated with congenital clubfoot and reduced penetrance in females.
Peterson J F, et al.
American Journal of Medical Genetics. Part A, 164(2), 364-369 (2014)
Rupak Datta et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(9), 3437-3442 (2009-02-13)
Missense mutations in the carbonic anhydrase IV (CA IV) gene have been identified in patients with an autosomal dominant form of retinitis pigmentosa (RP17). We used two transient expression systems to investigate the molecular mechanism by which the newly identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service