Skip to Content
Merck
All Photos(1)

Key Documents

SML1137

Sigma-Aldrich

ELN484228

≥98% (HPLC)

Synonym(s):

4′-Fluorobenzenesulfonanilide, N-(4-Fluorophenyl)-benzenesulfonamide, N-(4-Fluorophenyl)benzenesulfonamide, N-(p-Fluorophenyl)benzenesulfonamide, NSC 164389

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10FNO2S
CAS Number:
Molecular Weight:
251.28
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=S(C1=CC=CC=C1)(NC2=CC=C(F)C=C2)=O

InChI

1S/C12H10FNO2S/c13-10-6-8-11(9-7-10)14-17(15,16)12-4-2-1-3-5-12/h1-9,14H

InChI key

AZORQGXJAXEIGC-UHFFFAOYSA-N

Biochem/physiol Actions

ELN484228 is a cell penetrant α-synuclein ligand that rescues of α-synuclein induced disruption of vesicle trafficking and dopaminergic neuronal loss and neurite retraction. ELN484228 reverses a-synuclein induced impairment of phagocytosis and protects dopaminergic neurons against the toxic effects of α-synuclein A53T over-expression.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service