Skip to Content
Merck
All Photos(1)

Documents

SML0493

Sigma-Aldrich

T16Ainh-A01

≥95% (HPLC)

Synonym(s):

2-[(5-Ethyl-1,6-dihydro-4-methyl-6-oxo-2-pyrimidinyl)thio]-N-[4-(4-methoxyphenyl)-2-thiazolyl]-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N4O3S2
CAS Number:
Molecular Weight:
416.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

CCc1c(C)nc(SCC(=O)Nc2nc(cs2)-c3ccc(OC)cc3)nc1O

InChI

1S/C19H20N4O3S2/c1-4-14-11(2)20-18(23-17(14)25)28-10-16(24)22-19-21-15(9-27-19)12-5-7-13(26-3)8-6-12/h5-9H,4,10H2,1-3H3,(H,20,23,25)(H,21,22,24)

InChI key

QSIYTNYMBWYHAA-UHFFFAOYSA-N

Application

T16Ainh-A01 has been used as an inhibitor of transmembrane protein 16A (TMEM16A).

Biochem/physiol Actions

T16Ainh-A01 is a selective TMEM16A calcium-activated chloride channel inhibitor that strongly inhibits chloride current in salivary gland cells. T16Ainh-A01 poorly inhibited total CaCC current in intestinal and airway epithelial cells, but blocked mainly an initial, agonist-stimulated transient chloride current. T16Ainh-A01 does not alter cytoplasmic calcium and has little effect on CFTR Cl- conductance.

Features and Benefits

This compound is featured on the Chloride Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TMEM16A regulates portal vein smooth muscle cell proliferation in portal hypertension.
Zeng X, et al.
Experimental and Therapeutic Medicine, 15(1), 1062-1068 (2018)
Jinyu Zhang et al.
International journal of molecular sciences, 23(22) (2022-11-27)
Dry eye disease (DED) is a multifactorial disease with an incidence of approximately 50% worldwide. DED seriously affects quality of life and work. The prevalence of environmental DED (eDED) ranges from 35 to 48%. Conjunctival fluid secretion dysfunction may be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service