Skip to Content
Merck
All Photos(1)

Key Documents

SML0034

Sigma-Aldrich

1-EBIO

≥98% (HPLC)

Synonym(s):

1-Ethyl-1,3-dihydro-2H-benzimidazol-2-one; 1-Ethylbenzimidazolinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N2O
CAS Number:
Molecular Weight:
162.19
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: ≥20 mg/mL

storage temp.

room temp

SMILES string

CCN1C(=O)Nc2ccccc12

InChI

1S/C9H10N2O/c1-2-11-8-6-4-3-5-7(8)10-9(11)12/h3-6H,2H2,1H3,(H,10,12)

InChI key

CXUCKELNYMZTRT-UHFFFAOYSA-N

Application

1-EBIO was used to study the electrical activity modulated by Ca+2-activated K+ channels in rat central neurons. The effect of 1-EBIO on human intermediate conductance channels was studied.

Biochem/physiol Actions

1-EBIO is an activator of epithelial Ca2+-activated K+ channel also known as the intermediate-conductance Ca(2+)-activated K(+) channel (IK-1) or KCa3.1.
1-EBIO potentiates the secretion of CFTR-mediated Cl ion secretion by activating the basolateral KCNN4 K+ channels in native human colonic tissues. It stimulates the apical Cl channels and Ca+2-dependent basolateral K+ channels to stimulate fluid transport in lacrimal glands.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Audrey E Padula et al.
Translational psychiatry, 10(1), 414-414 (2020-11-29)
Mood disorders are often comorbid with alcohol use disorder (AUD) and play a considerable role in the development and maintenance of alcohol dependence and relapse. Because of this high comorbidity, it is necessary to determine shared and unique genetic factors
Eva K Roth et al.
PloS one, 6(8), e24445-e24445 (2011-09-13)
The identification of strategies to improve mutant CFTR function remains a key priority in the development of new treatments for cystic fibrosis (CF). Previous studies demonstrated that the K⁺ channel opener 1-ethyl-2-benzimidazolone (1-EBIO) potentiates CFTR-mediated Cl⁻ secretion in cultured cells
F Woodward Hopf et al.
Neuron, 65(5), 682-694 (2010-03-13)
The cellular mechanisms underlying pathological alcohol seeking remain poorly understood. Here, we show an enhancement of nucleus accumbens (NAcb) core action potential firing ex vivo after protracted abstinence from alcohol but not sucrose self-administration. Increased firing is associated with reduced
Lawrence J Alvarez et al.
American journal of physiology. Cell physiology, 289(1), C138-C147 (2005-02-11)
Experiments were conducted to determine whether the Cl- secretagogue, 1-ethyl-2-benzimidazolinone (EBIO), stimulates Cl- transport in the rabbit conjunctival epithelium. For this study, epithelia were isolated in an Ussing-type chamber under short-circuit conditions. The effects of EBIO on the short-circuit current
Patrick J Mulholland et al.
Biological psychiatry, 69(7), 625-632 (2010-11-09)
Small conductance calcium-activated potassium type 2 channels (SK2) control excitability and contribute to plasticity by reducing excitatory postsynaptic potentials. Recent evidence suggests that SK2 channels form a calcium-dependent negative-feedback loop with synaptic N-methyl-D-aspartate (NMDA) receptors. Addiction to alcohol and other

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service