Skip to Content
Merck
All Photos(1)

Key Documents

SMB00072

Sigma-Aldrich

Salidroside

≥95% (LC/MS-ELSD)

Synonym(s):

2-(4-Hydroxyphenyl)ethyl β−D-glucopyranoside, p-Hydroxyphenethyl glucopyranoside, Rhodioloside, Rhodosin, Tyrosol a-(β-D-glucopyranoside)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20O7
CAS Number:
Molecular Weight:
300.30
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](OCCc2ccc(O)cc2)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C14H20O7/c15-7-10-11(17)12(18)13(19)14(21-10)20-6-5-8-1-3-9(16)4-2-8/h1-4,10-19H,5-7H2/t10-,11-,12+,13-,14-/m1/s1

InChI key

ILRCGYURZSFMEG-RKQHYHRCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural product derived from plant source.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng Qian et al.
RNA biology, 16(10), 1461-1470 (2019-06-30)
Diabetic retinopathy (DR) is a complication of diabetes leading cause of blindness in adults. Salidroside (SAL) is a main ingredient from Rhodiola rosea L., has been reported to have a beneficial protection on vascular function. However, whether SAL is a
Lanqing Ma et al.
Sheng wu gong cheng xue bao = Chinese journal of biotechnology, 28(3), 282-294 (2012-06-21)
Salidroside, the 8-O-beta-D-glucoside of tyrosol, is a novel adaptogenic drug extracted from the medicinal plant Rhodiola sachalinensis A. Bor. Due to the scarcity of R. sachalinensis and its low yield of salidroside, there is great interest in enhancing the production
Wenfang Lai et al.
Neurotoxicity research, 28(2), 108-121 (2015-04-26)
Salidroside exhibits anti-inflammatory, anti-oxidative, and anti-apoptotic properties. To identify whether salidroside might be a candidate for treating ischemic stroke, we investigated the effects of salidroside or vehicle, given daily for 6 days, after middle cerebral artery occlusion (MCAO) for 2
A Alameddine et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 66(2), 249-257 (2015-04-24)
Many factors, including hyperglycemia, hypertension, obesity, dyslipidemia, and a sedentary lifestyle, contribute to a high prevalence of cardiovascular disease. Specific vascular impairment treatments in the context of diabetes and vascular risk need to be improved. Salidroside is the primary active
Xiu-Wen Wu et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 38(21), 3656-3660 (2014-02-06)
Salidroside, one of the active components of Rhodiola plants, is a phenolic glycoside with significant biological activities. The investigation and development of alternative production approaches of salidroside is of high academic and application values due to the limited resource of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service