SBCT06
SB-HEK293-Mock-CTRL
Negative control membrane for HEK293 transporter membranes
Synonym(s):
Negative control HEK293 human membrane, recombinant, expressed in human HEK293 cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
Recommended Products
recombinant
expressed in human cells
form
liquid
concentration
5.0 mg/mL
color
off-white
shipped in
dry ice
storage temp.
−70°C
General description
Membrane Preparations for Vesicular Transport Assays (VT) are suitable for general drug-efflux transporter interaction studies. Both substrate and inhibitor interactions can be assessed using vesicles. Membrane Preparations for ATPase Assays are suitable for drug-efflux transporter interaction studies based on ATPase activity determination, and could be used for differentiation between transporter substrates and inhibitors. Control Membranes with no-, or significantly lower transporter activity are available for both Vesicular Transport and ATPase Assays.
Application
Control Membranes with no-, or significantly lower transporter activity provide appropriate negative controls for both Vesicular Transport and ATPase assays. Refer to individual Vesicular Transport or ATPase assay product detail pages for appropriate control membranes needed for a specific assay.
Packaging
250 μL
Physical form
Supplied as frozen membrane vesicles, containing 5 mg/ml membrane protein, labeled with volume, catalog number (transporter) and date of production.
Legal Information
Distributed for SOLVO Biotechnology, Inc.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service