Skip to Content
Merck
All Photos(1)

Key Documents

SAB4503260

Sigma-Aldrich

Anti-Dysferlin, antibody produced in rabbit

affinity isolated antibody

Synonym(s):

dysferlin, limb girdle muscular dystrophy 2B (autosomal recessive)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 237 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DYSF(8291)

General description

Dysferlin (DYSF) is a large transmembrane protein that belongs to the ferlin family of Ca2+-dependent phospholipid-binding proteins. It is expressed in skeletal and cardiac muscles. DYSF gene is mapped to human chromosome 2p13. Anti-Dysferlin antibody detects endogenous levels of total dysferlin protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Dysferlin.

Immunogen Range: 1981-2030

Application

Anti-Dysferlin, antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Dysferlin (DYSF) plays a key role in membrane repair, vesicle trafficking, and Ttubule structure. Lack of DYSF protein expression in skeletal muscles leads to dysferlinopathy. Mutations in the DYSF gene are associated with Miyoshi myopathy, limb-girdle muscular dystrophy type 2B (LGMD2B), and distal myopathy with anterior tibial onset (DMAT).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Tang et al.
Neuropathology : official journal of the Japanese Society of Neuropathology (2018-05-26)
Dysferlinopathy, a progressive muscular dystrophy, results from mutations in the Dysferlin gene (DYSF, MIM*603009). Traditional diagnosis relies on the reduction or absence of dysferlin. However, altered dysferlin has been observed in other myopathies, leading to a precise diagnosis through molecular
Functional recovery of a novel knock-in mouse model of dysferlinopathy by readthrough of nonsense mutation
Seo K, et al.
Molecular therapy. Methods & clinical development (2021)
Kyowon Seo et al.
Molecular therapy. Methods & clinical development, 21, 702-709 (2021-06-19)
Biallelic mutations in the dysferlin gene cause limb-girdle muscular dystrophy 2B or Miyoshi distal myopathy. We found that nonsense mutations are the most common mutation type among Korean patients with dysferlinopathy; more than half of the patients have at least
Janice A Dominov et al.
Annals of clinical and translational neurology, 6(4), 642-654 (2019-04-26)
Dysferlin is a large transmembrane protein that functions in critical processes of membrane repair and vesicle fusion. Dysferlin-deficiency due to mutations in the dysferlin gene leads to muscular dystrophy (Miyoshi myopathy (MM), limb girdle muscular dystrophy type 2B (LGMD2B), distal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service