Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500863

Sigma-Aldrich

Anti-PRRT2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 43 kDa

species reactivity

rat, mouse, human

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRRT2(112476)

General description

Proline-rich transmembrane protein 2 (PRRT2) has two transmembrane domains. PRRT2 is highly expressed in nervous system and spinal cord and less expressed in heart, lung, kidney and skin. In human chromosome, the gene PRRT2 is localized on 16p11.2.

Biochem/physiol Actions

Prrt2 is found in the pre- and post- synaptic membranes and is associated with Synaptosomal nerve-associated protein 25 (SNAP25). Prrt2 promotes neurotransmitter release and glutamine signalling. Mutations in PRRT2 impairs glutamine release. Mutations in PRRT2 leads to delay in neuronal migration and synaptic loss. Truncating mutations in PRRT2 causes paroxysmal kinesigenic dyskinesia. Mutations in PRRT2 also causes epilepsy and mental retardation. Mutation of PRRT2 in children causes infantile convulsions and choreoathetosis (ICCA) syndrome and in adolescent leads to onset-movement disorder.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500863.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exome sequencing identifies truncating mutations in PRRT2 that cause paroxysmal kinesigenic dyskinesia
Chen WJ, et al.
Nature Genetics, 43(12), 1252-1252 (2011)
PRRT2 mutant leads to dysfunction of glutamate signaling
Li M, et al.
International Journal of Molecular Sciences, 16(5), 9134-9151 (2015)
PRRT2 mutations lead to neuronal dysfunction and neurodevelopmental defects
Liu YT, et al.
Oncotarget, 7(26), 39184-39184 (2016)
Sarah E Heron et al.
American journal of human genetics, 90(1), 152-160 (2012-01-17)
Benign familial infantile epilepsy (BFIE) is a self-limited seizure disorder that occurs in infancy and has autosomal-dominant inheritance. We have identified heterozygous mutations in PRRT2, which encodes proline-rich transmembrane protein 2, in 14 of 17 families (82%) affected by BFIE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service