Skip to Content
Merck
All Photos(2)

Key Documents

SAB3500119

Sigma-Aldrich

Anti-OCIAD2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution, ab2

Synonym(s):

Anti-OCIA domain-containing protein 2, Anti-Ovarian cancer immuno-reactive antigen domain-containing protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... OCIAD2(132299)

Immunogen

OCIAD2 antibody was raised against a 17 amino acid peptide from near the amino terminus of human OCIAD2.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

OCIAD2 was identified by its sequence similarity with OCIAD1, and together OCIAD1 and OCIAD2 form the OCIA domain family. OCIAD2 mRNA was found to be expressed at higher levels in invasive adenocarcinoma mixed subtype with bronchioloalveolar carcinoma component (BAC) of the lung. Loss of OCIAD2 expression was significantly correlated with lymphatic invasion, blood vessel invasion, and lymph node metastasis, indicating that OCIAD2 may play a role in cell adhesion and prevention of cell migration. While the function of OCIAD2 is still unknown, its expression in adenocarcinoma with BAC component is significantly associated with a favorable prognosis and may serve as a marker for selecting tumors that are treatable by limited surgery.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500119.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rochelle C J D'Souza et al.
Science signaling, 7(335), rs5-rs5 (2014-07-25)
Transforming growth factor-β (TGF-β) signaling promotes cell motility by inducing epithelial-to-mesenchymal transitions (EMTs) in normal physiology and development, as well as in pathological conditions, such as cancer. We performed a time-resolved analysis of the proteomic and phosphoproteomic changes of cultured
Time-resolved dissection of early phosphoproteome and ensuing proteome changes in response to TGF-?.
D'Souza RC, Knittle AM, Nagaraj N, et al.
Science Signaling, 7(335), rs5-rs5 (2014)
Jonghee Han et al.
Cellular and molecular life sciences : CMLS, 71(13), 2561-2576 (2013-11-26)
The gamma (γ)-secretase holoenzyme is composed of four core proteins and cleaves APP to generate amyloid beta (Aβ), a key molecule that causes major neurotoxicity during the early stage of Alzheimer's disease (AD). However, despite its important role in Aβ

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service