Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300081

Sigma-Aldrich

Anti-LRP1 (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-α-2-macroglobulin receptor, Anti-A2MR, Anti-Apolipoprotein E receptor, Anti-LRP, Anti-Low-density lipoprotein receptor-related protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LRP1(4035)

General description

LRP1 (low density receptor protein 1) gene codes for a protein belonging to LDL receptor family. Lrp1 is known to be the largest mammalian endocytic receptor. The encoded protein is cysteine-rich and it is highly glycosylated. It is predominantly expressed in vascular smooth muscle cells, neurons, astrocytes, macrophages, hepatocytes, fibroblasts and malignant cells. The LDLRs (low-density lipoprotein receptors) have five domains namely repeated cysteine-rich regions of ligand-binding type and epidermal growth factor receptor-like repeats, YWTD (tyrosin, typrtophan, Threonine, aspartic) domains, a transmembrane domain, and a cytoplasmic tail that holds about three NPXY (asparagine-proline-X-tyrosine) motifs.

Immunogen

LRP1 (4520-4554)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human LRP1.

Biochem/physiol Actions

LRP1 functions as a scavenger by internalizing several plasma components and also serves as a signaling molecule. Lrp1 is responsible for the transport of β-amyloid (Aβ) across the blood brain barrier, thereby contributing to the pathogenesis of Alzheimer′s disease. Lrp1 is normally associated with the cell membrane, but proteolytic cleavage of the cell results in its soluble form. Therefore, it loses its ability to endocytose and transport ligands. LRP1 is associated with a number of signaling pathways, such as mitogen-activated protein kinase (MAPK), serine/threonine protein kinase (AKT), insulin receptor (IR), extracellular signal-regulated kinase (ERK), and c-jun N-terminal kinase (JNK) pathways.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia A Young et al.
The Journal of biological chemistry, 291(50), 26035-26044 (2016-10-31)
Hemophilia A is a bleeding disorder caused by a deficiency in coagulation factor VIII (fVIII) that affects 1 in 5,000 males. Current prophylactic replacement therapy, although effective, is difficult to maintain due to the cost and frequency of injections. Hepatic
Dong-Chuan Guo et al.
American journal of human genetics, 99(3), 762-769 (2016-08-30)
Acute aortic dissections are a preventable cause of sudden death if individuals at risk are identified and surgically repaired in a non-emergency setting. Although mutations in single genes can be used to identify at-risk individuals, the majority of dissection case
Peipei Xing et al.
Chinese journal of cancer, 35, 6-6 (2016-01-08)
Low-density lipoprotein receptor-related protein 1 (LRP1, also known as CD91), a multifunctional endocytic and cell signaling receptor, is widely expressed on the surface of multiple cell types such as hepatocytes, fibroblasts, neurons, astrocytes, macrophages, smooth muscle cells, and malignant cells.
B Shackleton et al.
Fluids and barriers of the CNS, 13(1), 14-14 (2016-08-10)
Transport across the blood-brain barrier (BBB) is an important mediator of beta-amyloid (Aβ) accumulation in the brain and a contributing factor in the pathogenesis of Alzheimer's disease (AD). One of the receptors responsible for the transport of Aβ in the
Camilla De Nardis et al.
The Journal of biological chemistry, 292(3), 912-924 (2016-12-14)
LDL receptor-related protein 1 (LRP1) is a highly modular protein and the largest known mammalian endocytic receptor. LRP1 binds and internalizes many plasma components, playing multiple crucial roles as a scavenger and signaling molecule. One major challenge to studying LRP1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service