Skip to Content
Merck
All Photos(1)

Documents

S8015

Sigma-Aldrich

SSC Buffer

for Northern and Southern blotting, powder blend

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319
NACRES:
NA.25

grade

for molecular biology

form

powder blend

pH

6.9-7.1 (20-25 °C, 20 ×)

foreign activity

DNAse, none detected
Endonuclease, none detected
Exonuclease, none detected
NICKase, none detected
RNAse, none detected

storage temp.

room temp

General description

SSC (Saline Sodium Citrate) Buffer 20x powder is a standard reagent in Southern and Northern hybridization procedures. It is used to control stringency of wash buffer for the washing steps after hybridization. SSC buffer powder should be reconstituted to the final indicated volume using molecular biology grade water.

Application

Suitable for use in washing buffer for Northern and Southern hybridization procedures.

Features and Benefits

  • Easy to reconstitute powder
  • Multi-purpose use in hybridizations

Components

SSC Buffer 20x contains 3M NaCl in 0.3M sodium citrate (pH 7.0).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Bogers et al.
European journal of cardio-thoracic surgery : official journal of the European Association for Cardio-thoracic Surgery, 2(3), 160-166 (1988-01-01)
To identify risk factors for hospital mortality after postinfarction left ventricular aneurysmectomy (PILVA), 83 consecutive cases of PILVA were studied in a retrospective analysis. Hospital mortality was 14.4%. Most of the factors correlated with hospital mortality were associated with poor
Christèle Maison et al.
Nature genetics, 43(3), 220-227 (2011-02-15)
HP1 enrichment at pericentric heterochromatin is considered important for centromere function. Although HP1 binding to H3K9me3 can explain its accumulation at pericentric heterochromatin, how it is initially targeted there remains unclear. Here, in mouse cells, we reveal the presence of
Aline V Probst et al.
Developmental cell, 19(4), 625-638 (2010-10-19)
At the time of fertilization, the paternal genome lacks the typical configuration and marks characteristic of pericentric heterochromatin. It is thus essential to understand the dynamics of this region during early development, its importance during that time period and how

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service