Skip to Content
Merck
All Photos(1)

Documents

S1638

Millipore

Streptavidin−Agarose from Streptomyces avidinii

buffered aqueous suspension

Synonym(s):

streptavidin agarose beads, streptavidin agarose resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

form

buffered aqueous suspension

extent of labeling

≥1 mg per mL

technique(s)

affinity chromatography: suitable

matrix

4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

7 atoms

capacity

≥15 μg/mL binding capacity (biotin)

storage temp.

2-8°C

Application

Streptavidin−agarose from Streptomyces avidinii has been used:
  • to pull down biotinylated cell surface proteins during the quantification of plasma membrane transforming growth factor β (TGFβ) receptor II (TβRII) and Tβ
  • RII internalization
  • in biotinylated miRNA pull-down assay; as secondary antibodies in immunoprecipitation

Streptavidin-agarose is used in protein chromatography, affinity chromatography, and recombinant protein expression and analysis. Streptavidin-agarose has been used to study the oriented immobilization of the tobacco etch virus protease for the cleavage of fusion proteins. Streptavidin-agarose has also been used to develop a method for screening triplex DNA binders from natural plant extracts.
Used for the purification of biotin containing proteins or DNA binding proteins

Biochem/physiol Actions

Streptavidin is a homotetrameric protein, isolated from Streptomyces avidinii, which, like avidin, has a high affinity for biotin. Streptavidin is slightly anionic (pI ~ 5-6) and non-glycosylated. These properties contribute to its relatively low non-specific binding compared to egg white avidin. Streptavidin is also more resistant than avidin to dissociation into subunits by guanidinium chloride. Streptavidin-agarose can be used to immobilize or isolate various biotinylated macromolecules and complexes (proteins, antibodies, lectins, nucleic acids, receptors, and ligands). The inherent high-affinity streptavidin-biotin interaction requires harsh conditions to release biotinylated macromolecules. This feature makes streptavidin-agarose useful in a variety of affinity purification applications.

Physical form

Suspension in 0.01 M sodium phosphate, pH 7.2, containing 0.05 M NaCl and 0.02% sodium azide

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Gottschalk et al.
European journal of biochemistry, 267(23), 6875-6882 (2000-11-18)
Two cytochalasin B-binding states of the human red blood cell facilitative glucose transporter GLUT1 were studied, one exhibiting one cytochalasin B-binding site on every second GLUT1 monomer (state 1) and the other showing one site per monomer (state 2). Quantitative
Niusheng Xu et al.
Analytical chemistry, 84(5), 2562-2568 (2012-01-10)
A novel ligand fishing assay was established to screen triplex DNA binders from complicated samples by a combination of immobilization of triplex DNA on agarose beads and high-performance liquid chromatography-electrospray ionization-mass spectrometry (HPLC-ESI-MS). The biotinylated oligodeoxynucleotides were first bound to
Behrad Derakhshan et al.
Nature protocols, 2(7), 1685-1691 (2007-07-21)
Covalent addition of nitric oxide (NO) to Cys-sulfur in proteins, or S-nitrosylation, plays pervasive roles in the physiological and pathophysiological modulation of mammalian protein functions. Knowledge of the specific protein Cys residues that undergo NO addition in different biological settings
O Litzka et al.
European journal of biochemistry, 251(3), 758-767 (1998-03-07)
In Aspergillus nidulans, a DNA-binding complex, PENR1, was shown to bind to two CCAAT-box-containing DNA elements located in the promoter regions of the bidirectionally oriented penicillin biosynthesis genes acvA and ipnA, and of the aat promoter. Here, partial purification of
Jonathan A Roberts et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 27(15), 4072-4082 (2007-04-13)
P2X receptors for extracellular ATP are a distinct family of ligand-gated cation channels involved in physiological processes ranging from synaptic transmission to muscle contraction. Common ATP binding motifs are absent from P2X receptors, and the extent of the agonist binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service