RAB0659
Human FOLR1 / Folate Receptor Alpha ELISA Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.84
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type cell culture supernatant(s)
sample type plasma
sample type serum
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.1 ng/mL
standard curve range: 0.102-25 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... FOLR1(2348)
General description
The antibody pair provided in this kit recognizes human Folate Receptor Alpha.
The folate receptor 1 (FOLR1) gene is mapped to human chromosome 11q13.4. The gene codes for a glycosylphosphatidylinositol protein that is attached to cell membranes. Folr1 is expressed in placenta, kidney tubules and choroid plexus.
The folate receptor 1 (FOLR1) gene is mapped to human chromosome 11q13.4. The gene codes for a glycosylphosphatidylinositol protein that is attached to cell membranes. Folr1 is expressed in placenta, kidney tubules and choroid plexus.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Biochem/physiol Actions
Folate receptor 1 (FOLR1) plays a vital role in folate transport across the blood–brain barrier through the choroid plexus. It is also essential for neural tube closure during embryogenesis. Mutation in the gene leads to the development of cerebral folate deficiency in a few families. Overexpression of the gene has been observed in some epithelial malignancies including medulloblastoma, ovarian, lung and breast carcinomas. FOLR1 acts as a potential biomarker and therapeutic target for medulloblastoma.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Congenital null mutations of the FOLR1 gene: a progressive neurologic disease and its treatment.
Cario H, et al.
Neurology, 73(24), 2127-2129 (2009)
Update and new concepts in vitamin responsive disorders of folate transport and metabolism.
Watkins D and Rosenblatt DS
Journal of inherited metabolic disease, 35(4), 665-670 (2012)
Differential expression of folate receptor 1 in medulloblastoma and the correlation with clinicopathological characters and target therapeutic potential.
Liu H, et al.
Oncotarget, 8(14), 23048-23048 (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service