Skip to Content
Merck
All Photos(1)

Key Documents

R886N-10

Sigma-Aldrich

Rat Hippocampal Neurons: RHiN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

rat brain (hippocampus)

Quality Level

packaging

pkg of 1,000,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

morphology

Adherent. When cultured under the recommended conditions, cells arborize and form complex neurite network in one week.

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.


Rat Hippocampal Neurons are derived from hippocampi of day 18 embryonic Sprague Dawley rat brains. Cryopreserved immediately after isolation. >1,000,000 cells in Serum-free Freezing Medium.

Cell Line Origin

Brain

Application

Basic research on neuroscience, neurodevelopment, neuronal plasticity, memory formation, navigation, Alzheimer′s disease, stress and depression, epilepsy, and Schizophrenia, disease mechanisms, pathophysiologies, drug screening and development, toxicity testing, immunostaining, live cell imaging, co-culturing, electrophysiology.

Components

Serum-free Freezing Medium.

Quality

Each lot was tested for proper morphology, Stain positive for β III-Tubulin, Negative for HIV, Hepatitis B, Hepatitis C, mycoplasma, bacteria, and fungi.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service