Skip to Content
Merck
All Photos(1)

Key Documents

QR0200

Sigma-Aldrich

Quantitative RT-PCR ReadyMix

One step RT-qPCR for probe-based methods, MMLV & hot-start Taq

Synonym(s):

1-step RT-qPCR mix, Quantitative real-time PCR master mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

Quality Level

usage

sufficient for 250 reactions

feature

dNTPs included
hotstart

technique(s)

RT-qPCR: suitable

color

colorless

input

purified RNA

compatibility

ABI 5700
ABI 7000
ABI 7300
ABI 7500 Fast
ABI 7500
ABI 7700
ABI 7900 HT
ABI 7900 HT Fast
ABI 7900
ABI StepOne
ABI StepOnePlus
ABI ViiA 7
Bio-Rad CFX384
Bio-Rad CFX96
Bio-Rad MJ Chromo4
Bio-Rad MJ Opticon 2
Bio-Rad MJ Opticon
Bio-Rad MiniOpticon
Bio-Rad MyiQ
Bio-Rad iCycler iQ
Bio-Rad iQ5
Cepheid SmartCycler
Eppendorf® Mastercycler ep realplex2 s
Eppendorf® Mastercycler ep realplex
Illumina Eco qPCR
Qiagen Corbett Rotor-Gene 3000
Qiagen Corbett Rotor-Gene 6000
Qiagen Corbett Rotor-Gene Q
Roche LightCycler 480
Strategene Mx3000P
Strategene Mx3005P
Strategene Mx4000

detection method

probe-based

shipped in

wet ice

storage temp.

−20°C

General description

Quantitative RT-PCR ReadyMix combines Moloney Murine Leukemia Virus Reverse Transcriptase (M-MLV RT) and JumpStart Taq DNA polymerase in a one-step RT-PCR kit designed for the measurement of gene expression. This convenient 2X master mix includes M-MLV RT, JumpStart Taq DNA polymerase, 99% pure deoxynucleotides, buffer, glass passivator, and stabilizers. The JumpStart Taq DNA polymerase is an antibody-inactivated hot-start enzyme. Once the reaction temperature reaches 70°C, the DNA polymerase-antibody complex dissociates and Taq DNA polymerase activity is restored. This antibody-enzyme complex allows for easy and convenient set-up with less contamination risk than manual hot-start techniques. This kit has been formulated for fluorogenic hybridization probe-based detection methods.

Application

Quantitative RT-PCR ReadyMix has been used in the quantification of messenger RNA (mRNA) levels of specific expressed genes by quantitative reverse-transcription polymerase chain reaction (RT-qPCR).

Features and Benefits

  • The master mix allows consistency and reproducibility from one reaction to the next
  • Reduced risk of contamination from multiple pipetting steps
  • Reduced set-up time as compared to manual or wax Hot Start methods
  • JumpStartTaq Polymerase reduces primer-dimer and non-specific product formation
  • Broad instrument compatibility
  • Includes a separate ROX reference dye vial for reaction normalization

Packaging

1 kit sufficient for 250 reactions at 20 μL each or for 100 reactions at 50 μL each.

Legal Information

This product is for research use only.
Eppendorf is a registered trademark of Eppendorf AG
JumpStart is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • Probe Based qRT-PCR ReadyMix 2 X

  • Moloney Murine Leukemia Viral Reverse Transcriptase (M-MLV RT) 5000 U

Kit Components Also Available Separately

Product No.
Description
SDS

  • P219210X PCR Buffer, Optimized for routine PCR with MgCl2 included 1.5 mL/vialSDS

  • M8787Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR 25 mMSDS

  • P219210X PCR Buffer, Optimized for routine PCR with MgCl2 included 100 XSDS

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation of Quantitative PCR Assays
Lovatt, A., et al.
BioPharm., 22-32 (2002)
An Integrated High-Throughput System for mRNA Purification and Quantitation for Use in Identifying Gene Knockdown by RNA Interference
Wang, et al.
JALA: Journal of the Association for Laboratory Automation, 11, 314-318 (2006)
A screen of shRNAs targeting tumor suppressor genes to identify factors involved in A549 paclitaxel sensitivity
Ji D, et al.
Oncology Reports, 18(6), 1499-1505 (2007)
S A Bustin
Journal of molecular endocrinology, 29(1), 23-39 (2002-08-30)
The fluorescence-based real-time reverse transcription PCR (RT-PCR) is widely used for the quantification of steady-state mRNA levels and is a critical tool for basic research, molecular medicine and biotechnology. Assays are easy to perform, capable of high throughput, and can
T B Morrison et al.
BioTechniques, 24(6), 954-958 (1998-06-19)
Continuous fluorescence observation of amplifying DNA allows rapid and accurate quantification of initial transcript copy number. A simple and generic method for monitoring product synthesis with the double-stranded DNA dye, SYBR Green I provides initial template copy number estimation limited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service