Skip to Content
Merck
All Photos(1)

Key Documents

PZ0315

Sigma-Aldrich

Rupintrivir

≥98% (HPLC)

Synonym(s):

AG-7088, AG7088, Ethyl (E,4S)-4-[[(2R,5S)-2-[(4-fluorophenyl)methyl]-6-methyl-5-[(5-methyl-1,2-oxazole-3-carbonyl)amino]-4-oxoheptanoyl]amino]-5-[(3S)-2-oxopyrrolidin-3-yl]pent-2-enoate, Ruprintrivir

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H39FN4O7
CAS Number:
Molecular Weight:
598.66
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +30 to +40°, c = 0.5 in chloroform-d

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

room temp

InChI

1S/C31H39FN4O7/c1-5-42-27(38)11-10-24(16-21-12-13-33-29(21)39)34-30(40)22(15-20-6-8-23(32)9-7-20)17-26(37)28(18(2)3)35-31(41)25-14-19(4)43-36-25/h6-11,14,18,21-22,24,28H,5,12-13,15-17H2,1-4H3,(H,33,39)(H,34,40)(H,35,41)/b11-10+/t21-,22+,24+,28-/m0/s1

InChI key

CAYJBRBGZBCZKO-BHGBQCOSSA-N

General description

Rupintrivir is an irreversible peptidomimetic inhibitor designed to target human rhinovirus (HRV). It was previously named as AG7088. It has been suggested that rupintrivir may be involved in vitro antiviral activity.

Biochem/physiol Actions

Rupintrivir (AG7088) is an irreversible human rhinovirus (HRV) 3C protease inhibitor, also found to inhibit enterovirus and norovirus.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daisuke Sano et al.
Food and environmental virology, 13(1), 84-92 (2021-01-05)
This study investigated the influence of viral interference on the detection of enteric viruses using the integrated cell culture (ICC)-PCR with a BGM cell line. It was possible to detect 102 plaque-forming units (PFU)/flask of enterovirus 71 (EV71) in spite
S L Binford et al.
Antimicrobial agents and chemotherapy, 49(2), 619-626 (2005-01-28)
The picornavirus 3C protease is required for the majority of proteolytic cleavages that occur during the viral life cycle. Comparisons of published amino acid sequences from 6 human rhinoviruses (HRV) and 20 human enteroviruses (HEV) show considerable variability in the
Jing Wang et al.
Journal of virology, 85(19), 10021-10030 (2011-08-05)
EV71 is the primary pathogenic cause of hand-foot-mouth disease (HFMD), but an effective antiviral drug currently is unavailable. Rupintrivir, an inhibitor against human rhinovirus (HRV), has potent antiviral activities against EV71. We determined the high-resolution crystal structures of the EV71
Kim S Robinson et al.
Science (New York, N.Y.), 370(6521) (2020-10-24)
Immune sensor proteins are critical to the function of the human innate immune system. The full repertoire of cognate triggers for human immune sensors is not fully understood. Here, we report that human NACHT, LRR, and PYD domains-containing protein 1
Chunlong Ma et al.
ACS infectious diseases, 6(8), 2260-2270 (2020-07-22)
Enterovirus D68 (EV-D68) is a re-emerging virus that causes moderate to severe respiratory diseases in children. In severe cases, EV-D68 infection can lead to neurological complications called acute flaccid myelitis (AFM). There is currently no antiviral or vaccine available for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service