Skip to Content
Merck
All Photos(2)

Key Documents

PZ0117

Sigma-Aldrich

PF-573228

≥95% (HPLC)

Synonym(s):

6-[4-(3-Methanesulfonyl-benzylamino)-5-trifluoromethyl-pyrimidin-2-ylamino]-3,4-dihydro-1H-quinolin-2-one, PF-228

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20F3N5O3S
CAS Number:
Molecular Weight:
491.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

CS(=O)(=O)c1cccc(CNc2nc(Nc3ccc4NC(=O)CCc4c3)ncc2C(F)(F)F)c1

InChI

1S/C22H20F3N5O3S/c1-34(32,33)16-4-2-3-13(9-16)11-26-20-17(22(23,24)25)12-27-21(30-20)28-15-6-7-18-14(10-15)5-8-19(31)29-18/h2-4,6-7,9-10,12H,5,8,11H2,1H3,(H,29,31)(H2,26,27,28,30)

InChI key

HESLKTSGTIBHJU-UHFFFAOYSA-N

Biochem/physiol Actions

PF-573228 is a focal adhesion kinase (FAK) inhibitor; Non-receptor tyrosine kinase inhibitor.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Fak page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debarati Mukherjee et al.
American journal of translational research, 14(2), 1220-1233 (2022-03-12)
Post-therapeutic relapse remains the biggest challenge to breast cancer management. The re-initiation of proliferation of dormant tumor cells in either metastatic or primary tumor location marks the final rate-limiting step of malignancy and mortality. The underlying molecular mechanisms remain poorly
Shwetha Narasimhan et al.
Cytoskeleton (Hoboken, N.J.), 79(9-11), 81-93 (2022-08-24)
Ventral stress fibers (VSFs) are contractile actin fibers dynamically attached to cell-matrix focal adhesions. VSFs are critical in cellular traction force production and migration. VSFs vary from randomly oriented short, thinner fibers to long, thick fibers that span along the
Patricia Costa et al.
PloS one, 8(9), e74659-e74659 (2013-09-17)
Cell invasion through extracellular matrix (ECM) is a hallmark of the metastatic cascade. Cancer cells require adhesion to surrounding tissues for efficient migration to occur, which is mediated through the integrin family of receptors. Alterations in expression levels of β1
Jianghong Wu et al.
OncoTargets and therapy, 12, 3743-3751 (2019-06-14)
Aim: To study the carcinogenetic mechanism of HOXB7 in gastric cancer (GC) remains. Methods: Two human GC cell lines - SGC7901 and SNU1 - were used for this study. SGC7901 cells were transfected with siRNA-HOXB7 (siHOXB7) to knock down HOXB7
Nilay Taneja et al.
Cytoskeleton (Hoboken, N.J.), 77(9), 342-350 (2020-09-05)
The coordinated generation of mechanical forces by cardiac myocytes is required for proper heart function. Myofibrils are the functional contractile units of force production within individual cardiac myocytes. At the molecular level, myosin motors form cross-bridges with actin filaments and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service