Skip to Content
Merck
All Photos(1)

Key Documents

P8124

Sigma-Aldrich

Pyoverdines

from Pseudomonas fluorescens, >90% (HPLC)

Synonym(s):

Pyoverdine Detection, Pyoverdine Protein, Siderophore Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

Pseudomonas fluorescens

Quality Level

Assay

>90% (HPLC)

form

powder

solubility

H2O: ~10 mg/mL

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Pyoverdines from Pseudomonas fluorescens has been used :
  • in tryptophan fluorescence quenching to determine its binding to neutrophil-gelatinase-associated lipocalin (NGAL)
  • to detect pyoverdine diffusion surrounding siderophore-conjugated monobactam (MB-1)-resistant colonies in chelexed, dialyzed Mueller-Hinton broth (CDMHB)
  • for synchronous fluorescence spectra of purified pyoverdine

Biochem/physiol Actions

Pyoverdines help in promoting plant growth by chelating iron and rendering it unavailable for plant pathogens. The ferri-pyoverdine complex acts as a signaling molecule inducing the production of secreted virulence factors in Pseudomonas aeruginosa.
Pyoverdines, also called pseuobactins and pyoverdins, are fluorescent siderophores that have high affinity for iron (1032 M-1), and are synthesized by fluorescent pseudomonads under iron-deficient growth conditions. Pyoverdines were shown to prevent iron toxicity produced by iron overload in hepatocyte cultures and effectively scavenges the hydroxyl and peroxyl radicals. Pyoverdines are effective in acquiring iron from transferrin and lactoferrin. Pyoverdines are also involved in the suppression of pythium-induced damping-off of tomato and promotion of growth in some higher plants.
Pyoverdines, also called pseuobactins and pyoverdins, are fluorescent siderophores that have high affinity for iron, and are synthesized by fluorescent pseudomonads under iron-deficient growth conditions.

Physical form

Supplied as a mixture containing mainly the succinic acid (MW=1161), 2-hydroxy glutaramide (MW=1190), and succinamide (MW=1160) forms of pyoverdines.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification of Pyoverdines of Pseudomonas fluorescens 2-79 by Copper-Chelate Chromatography.
Xiao R and Kisaalita WS
Applied and Environmental Microbiology, 61(11), 3769-3774 (1995)
Correction: Multicolor Whole-Cell Bacterial Sensing Using a Synchronous Fluorescence Spectroscopy-Based Approach.
PLoS ONE (2015)
Pyoverdine, the Major Siderophore in Pseudomonas aeruginosa, Evades NGAL Recognition.
Peek ME
Interdisciplinary perspectives on infectious diseases, 843509-843509 (2012)
Adaptation-based resistance to siderophore-conjugated antibacterial agents by Pseudomonas aeruginosa.
Tomaras AP
Antimicrobial Agents and Chemotherapy, 57(9), 4197-4207 (2013)
Mylene Vaillancourt et al.
PLoS pathogens, 19(4), e1010942-e1010942 (2023-04-08)
During chronic cystic fibrosis (CF) infections, evolved Pseudomonas aeruginosa antibiotic resistance is linked to increased pulmonary exacerbations, decreased lung function, and hospitalizations. However, the virulence mechanisms underlying worse outcomes caused by antibiotic resistant infections are poorly understood. Here, we investigated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service