Skip to Content
Merck
All Photos(1)

Documents

P7119

Sigma-Aldrich

PGO Enzyme Preparation

1 G capsules

Synonym(s):

Peroxidase - Glucose oxidase preparation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

form

powder

solubility

H2O: soluble capsule/100 mL at 25 °C, clear, colorless (pH 6.95-7.05)

storage temp.

2-8°C

General description

Each capsule contains 500 units of glucose oxidase (Aspergilus niger), 100 purpurogallin units of peroxidase (horseradish), and buffer salts. The reaction produces oxidized o-Dianisidine, which is brown. The intensity of the brown color measured at 425-475 nm is proportional to the original glucose concentration.

Application

PGO Enzymes preparation has been used:
  • in diet sampling and chemical analyses
  • to determine plasma and follicular fluid glucose levels
  • to measure glucose content in fractions of retrograded debranched rice starch

Biochem/physiol Actions

PGO Enzymes are used for the quantitative, enzymatic determination of glucose in aqueous solutions such as serum. The reactions are normally monitored at 425-475 nm utilizing o-dianisidine as a colorimetric substrate. Glucose oxidase methods eliminate the effects of interfering substances. Contents of one capsule includes 100 units peroxidase and 500 units glucose oxidase and buffer salts.

Suitability

Tested and found suitable for use in the quantitation of glucose.

Preparation Note

The PGO enzymes solution is prepared by adding the contents of 1 capsule of PGO enzymes to 100 mL of water in an amber bottle. Bottle should be inverted several times with gentle shaking to dissolve. PGO Enzymes solution should be stored at 2-8 °C. The solution is stable for up to 1 month unless turbidity develops, and for at least 6 months at –20 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joel I Jokinen et al.
Plants (Basel, Switzerland), 8(6) (2019-06-05)
Infection by holoparasitic plants typically causes decreases in host mass, thought to be primarily as a result of resource abstraction. Inverse relationships have been noted between the number of Orobanche spp. parasites infecting a host and their mass, suggesting that
P Piantoni et al.
Journal of dairy science, 98(5), 3323-3334 (2015-03-03)
Forty-eight multiparous cows were used in a randomized complete block design experiment with a 2×2 factorial arrangement of treatments to determine the interaction between a highly saturated free FA supplement (SFFA) and dietary forage NDF (fNDF) content on energy balance
Reza Salehi et al.
Reproductive biology and endocrinology : RB&E, 13, 69-69 (2015-07-04)
The objective was to determine the effect of prepartum diets supplemented with rolled canola seed (high in oleic acid) or sunflower seed (high in linoleic acid) on luteinizing hormone (LH) pulsatility and gonadotropin releasing hormone (GnRH)-induced LH release during early
Evaluation of triticale dried distillers grains with solubles as a substitute for barley grain and barley silage in feedlot finishing diets
K. T. Wierenga, T. A. McAllister, et al.
Journal of Animal Science, 88, 2018-3029 (2010)
Effects of reducing dietary starch content by replacing barley grain with wheat dried distillers grains plus solubles in dairy cow rations on ovarian function
Subramaniam E, et al.
Journal of Dairy Science, 99, 2762-2774 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service