Skip to Content
Merck
All Photos(1)

Documents

P0537

Sigma-Aldrich

1,2-Dilinoleoyl-sn-glycero-3-phosphocholine

≥99% (TLC), lyophilized powder

Synonym(s):

(7R,18Z,21Z)-4-Hydroxy-N,N,N-trimethyl-10-oxo-7-[[(9Z,12Z)-1-oxo-9,12-octadecadien-1-yl]oxy]-3,5,9-trioxa-4-phosphaheptacosa-18,21-dien-1-aminium 4-oxide, inner salt, 1,2-Di(cis-9,12-octadecadienoyl)-sn-glycero-3-phosphocholine, 1,2-Dilinoleoyl-sn-3-glycerophosphorylcholine, 3-sn-Phosphatidylcholine, 1,2-dilinoleoyl, L-α-Dilinoleoyl phosphatidylcholine, L-Dilinoleoyllecithin, Dilinoleoyl-L-α-phosphatidylcholine, PC(18:2(9Z,12Z)/18:2(9Z,12Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H80NO8P
CAS Number:
Molecular Weight:
782.08
Beilstein:
5212188
EC Number:
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥99% (TLC)

form

lyophilized powder

functional group

amine
ester

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCC/C=C\C/C=C\CCCCC)=O)=O

InChI

1S/C44H80NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h14-17,20-23,42H,6-13,18-19,24-41H2,1-5H3/b16-14-,17-15-,22-20-,23-21-/t42-/m1/s1

InChI key

FVXDQWZBHIXIEJ-LNDKUQBDSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged under Argon.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lars V Schäfer et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(4), 1343-1348 (2011-01-06)
Cell membranes are comprised of multicomponent lipid and protein mixtures that exhibit a complex partitioning behavior. Regions of structural and compositional heterogeneity play a major role in the sorting and self-assembly of proteins, and their clustering into higher-order oligomers. Here
Wei Liu et al.
Free radical biology & medicine, 50(1), 166-178 (2010-11-05)
We report herein that oxidation of a mitochondria-specific phospholipid tetralinoleoyl cardiolipin (L(4)CL) by cytochrome c and H(2)O(2) leads to the formation of 4-hydroxy-2-nonenal (4-HNE) via a novel chemical mechanism that involves cross-chain peroxyl radical addition and decomposition. As one of
Joakim P M Jämbeck et al.
The journal of physical chemistry. B, 116(10), 3164-3179 (2012-02-23)
An all-atomistic force field (FF) has been developed for fully saturated phospholipids. The parametrization has been largely based on high-level ab initio calculations in order to keep the empirical input to a minimum. Parameters for the lipid chains have been
Jeffery B Klauda et al.
The journal of physical chemistry. B, 114(23), 7830-7843 (2010-05-26)
A significant modification to the additive all-atom CHARMM lipid force field (FF) is developed and applied to phospholipid bilayers with both choline and ethanolamine containing head groups and with both saturated and unsaturated aliphatic chains. Motivated by the current CHARMM
Joel H Nyström et al.
Biophysical journal, 99(2), 526-533 (2010-07-21)
Cholesterol is distributed unevenly between different cellular membrane compartments, and the cholesterol content increases from the inner bilayers toward the plasma membrane. It has been suggested that this cholesterol gradient is important in the sorting of transmembrane proteins. Cholesterol has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service