P0102
PKCβII Peptide Inhibitor I trifluoroacetate salt
≥95% (HPLC), lyophilized powder
Synonym(s):
Protein Kinase C Beta II Peptide Inhibitor I trifluoroacetate salt
About This Item
Recommended Products
Assay
≥95% (HPLC)
form
lyophilized powder
color
white
solubility
H2O: >1 mg/mL
shipped in
wet ice
storage temp.
−20°C
SMILES string
OC(=O)C(F)(F)F.CCCCCCCCCCCCCC(=O)N[C@@H](CO)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(N)=O)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](Cc2c[nH]c3ccccc23)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H]([C@@H](C)O)C(O)=O
InChI
1S/C61H94N12O19.C2HF3O2/c1-5-6-7-8-9-10-11-12-13-14-15-22-49(78)65-45(33-74)58(88)68-41(28-34(2)3)55(85)71-44(31-48(63)77)60(90)73-27-18-21-46(73)59(89)67-39(23-25-50(79)80)53(83)69-42(29-36-32-64-38-20-17-16-19-37(36)38)56(86)70-43(30-47(62)76)57(87)66-40(24-26-51(81)82)54(84)72-52(35(4)75)61(91)92;3-2(4,5)1(6)7/h16-17,19-20,32,34-35,39-46,52,64,74-75H,5-15,18,21-31,33H2,1-4H3,(H2,62,76)(H2,63,77)(H,65,78)(H,66,87)(H,67,89)(H,68,88)(H,69,83)(H,70,86)(H,71,85)(H,72,84)(H,79,80)(H,81,82)(H,91,92);(H,6,7)/t35-,39+,40+,41+,42+,43+,44+,45+,46+,52+;/m1./s1
InChI key
FATKAQBIAPMYQT-YPNBOXGNSA-N
Amino Acid Sequence
Biochem/physiol Actions
Features and Benefits
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service