Skip to Content
Merck
All Photos(2)

Key Documents

MSP03

Sigma-Aldrich

Membrane Scaffold Protein 1D1(-)

recombinant, expressed in E. coli, buffered aqueous solution

Synonym(s):

Membrane Scaffold Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

recombinant

expressed in E. coli

Assay

≥90% (SDS-GE)

form

buffered aqueous solution

mol wt

22,043.9 Da

solubility

water: soluble

shipped in

ambient

storage temp.

−20°C

General description

Research area: Cell structure. Membrane Scaffold Protein 1D1 (MSP1D1) is a variant of MSP1 where the first eleven amino acids in the helix 1 portion are deleted of the original MSP1 sequence. It consists of an N-terminal histidine-tagged protein with a tobacco etch virus (TEV) protease cleavage site that is present between the protein sequence and histidine-tag. MSP1D1(–)variant of MSP1D1 lacks the histidine-tag and the TEV protease-recognition sequence.

Application

For guidelines on the use of this and other MSP′s to prepare Nanodiscs, please visit our Protocols for Membrane Scaffold Proteins and Nanodisc Formation page.

Biochem/physiol Actions

Nano-disctechnology is an approach that allows aqueous soluble membrane proteins in anative-like bilayer environment that enables the membrane proteins to remainactive and stable. The concept of this technology is derived from high-densitylipoprotein (HDL) particles and apolipoprotein, their primary proteincomponent. The nano-disc is a non-covalent structure consisting of MSP that isgenetically engineered to mimic the activity of apolipoprotein A1 (Apo-A1) andphospholipid bilayer. In the presence of synthetic phospholipids, MSP1D1 canself-assemble into discoidal nanoparticlesknown as nano-discs. These nano-discs are suitable for studying membraneproteins and are applicable in the field of medicine and biotechnology.

Legal Information

Nanodisc technology, and many of its uses, are covered by the following patents held by the University of Illinois.
  • 7,691,414 Membrane scaffold proteins
  • 7,662,410 Membrane scaffold proteins and embedded membrane proteins
  • 7,622,437 Tissue factor compositions and methods
  • 7,592,008 Membrane scaffold proteins
  • 7,575,763 Membrane scaffold proteins and tethered membrane proteins
  • 7,083,958 Membrane scaffold proteins
  • 7,048,949 Membrane scaffold proteins

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ramona Faas et al.
Biotechnology reports (Amsterdam, Netherlands), 17, 45-48 (2018-01-31)
Membrane scaffold proteins (MSPs) are synthetic derivatives of apolipoprotein A-I, a major protein component of human high-density lipoprotein complexes. The most common among these is the variant MSP1D1, which has been in the focus of research on membrane mimetics in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service