Skip to Content
Merck
All Photos(1)

Key Documents

MBD0006

Sigma-Aldrich

Inactivated Akkermansia muciniphila

Suitable for DNA extraction, PCR, sequencing, next generation sequencing, >10^8 bacteria/vial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500
NACRES:
NA.51

Quality Level

form

powder

concentration

>10^8 bacteria/vial

technique(s)

DNA extraction: suitable
DNA sequencing: suitable
PCR: suitable

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

Standardization of sample analysis is currently needed in microbiome genomics research workflow. Lack of standardization can lead to biases and errors in common processes during sample preparation and analysis such as sample amplification, sequencing and bioinformatics analyses.1 Inactivated Akkermansia muciniphila can serve as standard for benchmarking the performance along the workflow of microbiomics or meta-genomics analyses and as a tool to increase reproducibility and allow comparison of results obtained by different labs.
Akkermansia muciniphila is a gram negative, anaerobic, non motile, non spore forming and oval shaped bacterium. A. muciniphila inhabits the gastrointestinal tracts of more than 90% of adults and constitutes 1 to 4% of the fecal microbiota.2 A. muciniphila level was suggested to be a biomarker for a healthy intestin. 3
A. muciniphila degrades mucus and utilizes it as a carbon/nitrogen source. Consequently, the host produces additional mucus while the bacterium produces oligosaccharides and Short Chain Fatty Acids (SCFAs) that can be utilized by the host and trigger the immune system. An additional protective effect of the SCFA is stimulation of mucus-associated microbiota growth, that serves as a barrier against penetration of pathogens to intestinal cells. 2,4

Read here how to use our standards to ensure data integrity for your microbiome research.


Application

Inactivated bacteria are provided in at >10^8 bacteria/ml concentration in TE buffer pH 8.0.It is recommended to avoid freeze thaw cycles of this product.

Physical form

Liquid -The inactivated bacteria is provided at >10^8 bacteria/ml in TE buffer pH 8.0

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service