Skip to Content
Merck
All Photos(1)

Documents

M2389

Sigma-Aldrich

N6-Methyl-2′-deoxyadenosine

Synonym(s):

6mdA, N6-Me-dAdo, m6dA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15N5O3
CAS Number:
Molecular Weight:
265.27
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥98% (TLC)

Quality Level

form

powder

solubility

acetic acid: 49.00-51.00 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CNc1ncnc2n(cnc12)[C@H]3C[C@H](O)[C@@H](CO)O3

InChI

1S/C11H15N5O3/c1-12-10-9-11(14-4-13-10)16(5-15-9)8-2-6(18)7(3-17)19-8/h4-8,17-18H,2-3H2,1H3,(H,12,13,14)/t6-,7+,8+/m0/s1

InChI key

DYSDOYRQWBDGQQ-XLPZGREQSA-N

Application

N6-Methyl-2′-deoxyadenosine has been used as a standard to assess the levels of modified bases in genomic DNA by the ultra-high-performance liquid chromatography coupled with triple quadrupole mass spectrometry (UHPLC-QQQ-MS/MS) method. It has also been used to study its potential in promoting the proliferation oferythroid progenitor cells.

Biochem/physiol Actions

N6-Methyl-2′-deoxyadenosine (N6-Me-dAdo) is a precursor of N6-methyl 2′-deoxyadenosine 3′,5′-bisphosphate (N6MABP), a competitive and selective inhibitor of P2Y1 receptors. It is located at the transcription start site and plays a role in increased gene expression in Chlamydomonas reindardtii. N6-Me-dAdo augments the within transposable elements and is present at higher levels during the development of Drosophila. It is involved in the regulation of gene transcription, DNA repair, and replication in prokaryotes and protists. N6-Me-dAdo complemented with gene bodies might induce gene expression in human cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Boyer et al.
British journal of pharmacology, 124(1), 1-3 (1998-06-18)
The antagonist activity of N6-methyl 2'-deoxyadenosine 3',5'-bisphosphate (N6MABP) has been examined at the phospholipase C-coupled P2Y1 receptor of turkey erythrocyte membranes. N6MABP antagonized 2MeSATP-stimulated inositol phosphate hydrolysis with a potency approximately 20 fold greater than the previously studied parent molecule
Shengjie Li et al.
Frontiers in microbiology, 10, 1905-1905 (2019-09-10)
DNA methylation serves as a vital component of restriction-modification (R-M) systems in bacteria, where it plays a crucial role in defense against foreign DNA. Recent studies revealed that DNA methylation has a global impact on gene expression. Deinococcus radiodurans, an
J C Rogers et al.
The Plant journal : for cell and molecular biology, 7(2), 221-233 (1995-02-01)
Methylation of deoxycytosine residues in plant nuclear DNA at CpG dinucleotides is generally assumed to suppress transcription, while deoxyadenosine methylation on recombinant reporter gene constructs transiently introduced into plant cells increases expression by uncharacterized mechanisms. A particle bombardment transient expression
J J Solomon et al.
Chemico-biological interactions, 51(2), 167-190 (1984-09-15)
Reaction of the rodent carcinogen acrylonitrile (AN) at pH 5.0 and/or pH 7.0 for 10 and/or 40 days with 2'-deoxyadenosine (dAdo), 2'-deoxycytidine (dCyd), 2'-deoxyguanosine (dGuo), 2'-deoxyinosine (dIno), N6-methyl-2'-deoxyadenosine (N6-Me-dAdo) and thymidine (dThd) resulted in the formation of cyanoethyl and carboxyethyl
Mariangela Chisari et al.
Molecular neurobiology, 54(6), 4081-4093 (2016-06-20)
Astrocytes regulate neuronal activity and blood brain barrier through tiny plasma membrane branches or astrocytic processes (APs) making contact with synapses and brain vessels. Several transmitters released by astrocytes and exerting their action on several receptor classes expressed by astrocytes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service