Skip to Content
Merck
All Photos(1)

Key Documents

L4631

Sigma-Aldrich

Lysozyme from chicken egg white

For use as a marker in SDS-PAGE

Synonym(s):

Mucopeptide N-acetylmuramoylhydrolase, Muramidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

chicken egg white

Quality Level

form

powder

mol wt

single-chain 14.3 kDa

packaging

vial of 25 mg

UniProt accession no.

storage temp.

2-8°C

Gene Information

chicken ... LYZ(396218)

Looking for similar products? Visit Product Comparison Guide

Application

Enzyme breaks down the cell walls of bacteria; used to prepare spheroplasts.

Biochem/physiol Actions

Lysozyme hydrolyzes β(1→4) linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrin. Gram-positive cells are quite susceptible to this hydrolysis as their cell walls have a high proportion of peptidoglycan. Gram-negative bacteria are less susceptible due to the presence of an outer membrane and a lower proportion of peptidoglycan. However, these cells may be hydrolyzed in the presence of EDTA that chelates metal ions in the outer bacterial membrane.

The enzyme is active over a broad pH range (6.0 to 9.0). At pH 6.2, maximal activity is observed over a wider range of ionic strengths (0.02 to 0.100 M) than at pH 9.2 (0.01 to 0.06 M).

Unit Definition

One unit will lyse 0.6 μg of Micrococcus lysodeikticus per minute by turbidimetric detection at 600 nm when suspended in buffer at pH 6.2 at 25 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enav Bar Shira et al.
PloS one, 13(7), e0200393-e0200393 (2018-07-07)
Intestinal epithelial cells are multi-tasked cells that participate in digestion and absorption as well as in protection of the digestive tract. While information on the physiology and immune functions of intestinal epithelial cells in mammals is abundant, little is known
Anne Derbise et al.
The Journal of infectious diseases, 207(10), 1535-1543 (2013-02-14)
Yersinia pestis (the plague bacillus) and its ancestor, Yersinia pseudotuberculosis (which causes self-limited bowel disease), encode putative homologues of the periplasmic lysozyme inhibitor Ivy and the membrane-bound lysozyme inhibitor MliC. The involvement of both inhibitors in virulence remains subject to
Mariela G Del Giudice et al.
Infection and immunity, 81(3), 956-964 (2013-01-16)
Secretion of proteins in Gram-negative bacteria is a high-energy-consuming process that requires translocation across two membranes and a periplasmic space composed of a mesh-like layer, the peptidoglycan. To achieve this, bacteria have evolved complex secretion systems that cross these barriers
P F Zabrodskiĭ et al.
Eksperimental'naia i klinicheskaia farmakologiia, 75(11), 19-21 (2013-01-18)
It is established in experiments on noninbred rats that the use of imunofan (20 mg/kg daily) and polyoxidonium (150 mg/kg daily) for 7 days on the background of chronic intoxication with organophosphorus agent VX (0.01 LD50, single daily treatment for
Andrea Weeks et al.
Eye & contact lens, 39(2), 179-185 (2013-02-16)
This study was designed to assess the duration of hyaluronic acid (HA) release from model contact lens materials when HA was physically incorporated into the hydrogel during synthesis and to assess the effects of the HA release on lysozyme sorption.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service