Skip to Content
Merck
All Photos(1)

Key Documents

I7760

Sigma-Aldrich

IPL-41 Insect Medium

With ʟ-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Insect Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Product Name

IPL-41 Insect Medium, With L-glutamine and sodium bicarbonate., liquid, sterile-filtered, suitable for insect cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | insect: suitable

components

NaHCO3: 0.25 g/L

shipped in

ambient

storage temp.

2-8°C

Application

IPL-41 Insect Medium has been used in culturing silk worm cell lines.
IPL-41 is part of a series of media originally developed at the USDA Insect Pathology Laboratory by Weiss et al. for the large scale propagation of cells from the fall armyworm, Spodoptera frugiperda. The medium is primarily used for the growth and maintenance of cell lines derived from lepidopterans and the propagation of viruses in these cell lines. One of the most significant uses of the medium is the large scale culture of baculovirus infected Spodoptera cells. The expression of recombinant proteins is undertaken from Spodoptera cells utilizing the baculovirus expression system (BEVS). Attempts have also been made to obtain the expression of foreign genes in cells cultured in IPL-41 without serum supplementation.

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effective RNA interference in cultured silkworm cells mediated by overexpression of Caenorhabditis elegans SID-1
Mon H, et al.
RNA Biology, 9(1), 40-46 (2012)
Chromatin-induced spindle assembly plays an important role in metaphase congression of silkworm holocentric chromosomes
Mon H, et al.
Insect Biochemistry and Molecular Biology, 45(4), 40-50 (2014)
Masashi Iwanaga et al.
Journal of invertebrate pathology, 101(2), 124-129 (2009-05-23)
A new cell line, designated as NIAS-Boma-529b, was established from the larval fat bodies of Bombyx mandarina (B. mandarina), which is believed to be an ancestor of Bombyx mori (B. mori). This cell line has been cultured for approximately 150
Hiroaki Mon et al.
RNA biology, 9(1), 40-46 (2012-02-02)
RNA interference (RNAi) is a conserved mechanism that catalyzes sequence-specific gene silencing and has been used for loss-of-function genetic screens in many organisms. Here, we demonstrated that the expression of Caenorhabditis elegans SID-1 (CeSID-1) could trigger effective gene silencing in
Italo Delalibera et al.
Journal of invertebrate pathology, 84(2), 119-127 (2003-11-15)
The pathogenic fungus Neozygites tanajoae, one of the most efficient natural enemies of the cassava green mite (CGM) Mononychellus tanajoa in Brazil, was introduced experimentally in Benin in 1998/1999 for the control of CGM. Isolation methods and culture media for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service