Skip to Content
Merck
All Photos(1)

Documents

I5408

Sigma-Aldrich

EX-CELL® TiterHigh Medium

Animal-component free, liquid, sterile-filtered, suitable for insect cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41161501
NACRES:
NA.75

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | insect: suitable

storage temp.

2-8°C

Application

Animal component-free, protein-free formulation designed specifically for the Sf21 and Sf9 (Spodoptera frugiperda) insect cell lines

Features and Benefits

Designed to meet the needs of the biopharmaceutical industry, this animal component-free medium supports fast cell growth rates and high cell densities, while maintaining high cell viability and high recombinant protein production using the Baculovirus Expression Vector System (BEVS).

Components

This formulation includes inorganic salts, sodium bicarbonate, essential and non-essential amino acids, vitamins, yeast extract, a proprietary lipid formulation, trace elements, and other organic compounds. Glutamine is supplied in the form of an alanine-glutamine dipeptide. It does not contain phenol red, antibiotics, antimycotics, transferrin, and products of animal origin.

Caution

Do not freeze. Protect from light. Several components of the medium are light labile and should not be exposed to light for lengthy periods of time.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany
TiterHigh is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service