Skip to Content
Merck
All Photos(6)

Key Documents

HPA045827

Sigma-Aldrich

Anti-CD40LG antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD154, Anti-CD40L, Anti-HIGM1, Anti-IMD3, Anti-TNFSF5, Anti-TRAP, Anti-gp39, Anti-hCD40L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

LTVKRQGLYYIYAQVTFCSNREASSQAPFIASLCLKSPGRFERILLRAANTHSSAKPCGQQSIHLGGVFELQPGASVFVNVTDPSQVS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD40LG(959)

General description

The CD40 ligand (CD40L/CD40LG), a type II membrane protein, belongs to the tumor-necrosis factor (TNF) superfamily. It has a sandwich extracellular structure that consists of a β-sheet, α-helix loop, and a β-sheet. The CD40LG gene is located on human chromosome Xq26.3.

Immunogen

CD40 ligand recombinant protein epitope signature tag (PrEST)

Application

Anti-CD40LG antibody produced in rabbit has been used in multiparameter immunofluorescence histology (1:50).

Biochem/physiol Actions

The CD40 ligand (CD40L/CD40LG) plays a regulatory role in B-cell fate during various aforementioned checkpoints in B-cell development and differentiation. It is essential for the development of anti-tumor immunity. CD40L can be used as a cancer-related target in antibody-based therapy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78058

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P DiSanto et al.
Nature, 361(6412), 541-543 (1993-02-11)
Signalling for the B-cell immunoglobulin isotype switch requires T-cell-derived cytokines and T-B cell interaction, which operates primarily through the CD40 molecule on B cells with its ligand (CD40L) on activated T cells (reviewed in ref. 1). The CD40L is a
Raul Elgueta et al.
Immunological reviews, 229(1), 152-172 (2009-05-12)
During the generation of a successful adaptive immune response, multiple molecular signals are required. A primary signal is the binding of cognate antigen to an antigen receptor expressed by T and B lymphocytes. Multiple secondary signals involve the engagement of
Karlyn E Pollack et al.
Journal for immunotherapy of cancer, 8(1) (2020-05-01)
Immunogenicity of cancer vaccines is impacted by adjuvants and schedule, but systematic assessments of their effects have not been performed. Montanide ISA-51, an incomplete Freund's adjuvant (IFA), is used in many vaccine trials, but concerns have been raised about negative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service