Skip to Content
Merck
All Photos(4)

Key Documents

HPA028251

Sigma-Aldrich

Anti-FGFR4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD334, Anti-JTK2, Anti-fibroblast growth factor receptor 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

EEVELEPCLAPSLEQQEQELTVALGQPVRLCCGRAERGGHWYKEGSRLAPAGRVRGWRGRLEIASFLP

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FGFR4(2264)

General description

The gene FGFR4 (fibroblast growth factor receptor 4) is mapped to human chromosome 5q35.2. It belongs to the fibroblast growth factor receptor (FGFR) tyrosine kinase family of proteins. The protein has three extracellular immunoglobulin-type domains (D1-D3), a single-span transmembrane domain and an intracellular split tyrosine kinase domain.

Immunogen

fibroblast growth factor receptor 4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FGFRs (fibroblast growth factor receptors) control various cellular processes such as cell growth, differentiation, survival and migration. FGFR4 is an oncogene and is associated with tumor development. It is associated with colorectal cancer, ovarian cancer and hepatocellular carcinoma. It binds to FGFs (fibroblast growth factors), multifunctional proteins involved in angiogenesis, wound healing and embryonic development.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78001

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fibroblast growth factor receptor 4 protein expression and clinicopathological features in gastric cancer.
Chen H, et al.
World Journal of Gastroenterology, 21, 1838-1844 (2015)
Fibroblast growth factor receptor 4 polymorphism is associated with liver cirrhosis in hepatocarcinoma.
Sheu MJ, et al.
PLoS ONE, 10, e0122961-e0122961 (2015)
Evolution of the Fgf and Fgfr gene families.
Itoh N and Ornitz DM
Trends in Genetics, 20, 563-569 (2004)
FGFR4 Is a Potential Predictive Biomarker in Oral and Oropharyngeal Squamous Cell Carcinoma.
Koole K, et al.
Pathobiology, 82, 280-289 (2015)
Kelly J Soady et al.
Development (Cambridge, England), 144(20), 3777-3788 (2017-09-06)
PTPRB is a transmembrane protein tyrosine phosphatase known to regulate blood vessel remodelling and angiogenesis. Here, we demonstrate that PTPRB negatively regulates branching morphogenesis in the mouse mammary epithelium. We show that Ptprb is highly expressed in adult mammary stem

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service