Skip to Content
Merck
All Photos(2)

Key Documents

HPA025078

Sigma-Aldrich

Anti-LEMD3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BMP-3b, Anti-LEM domain containing 3, Anti-MAN1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

QGQAFHLDRRNSPPNSLTPCLKIRNMFDPVMEIGDQWHLAIQEAILEKCSDNDGIVHIAVDKNSREGCVYVKCLSPEYAGKAFKALHGSWFDGKLVTVKYLRLDRYHHRFPQALTSNTPLKPSNKHMNSMSHLRLRT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LEMD3(23592)

General description

The gene LEMD3 (LEM domain containing 3) is mapped to human chromosome 12q14. The encoded protein localizes in the inner nuclear membrane. It belongs to the LEM protein family. The protein has nucleoplasmic domains at the carboxy and amino terminals. One nucleoplasmic domain contains LEM motif and the other has binding sites for Smad2 (also referred to as MADH2 (mothers against decapentaplegic homolog 2)) and Smad3.

Immunogen

LEM domain containing 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

LEM proteins are mainly involved in gene regulation, chromatin arrangement and regulation of transcription factor. LEMD3 (LEM domain containing 3) interaction with Smad1/2 (also referred to as MADH (mothers against decapentaplegic homolog)) and Samd3 suppresses BMP (bone morphogenic protein) and TGF (transforming growth factor)-β signaling, respectively. It also plays an important role in the regulation of the clock protein, BMAL1 (brain and muscle ARNT-like 1). Absence of LEMD3 activity causes Buschke-Ollendorff syndrome and sclerosing bone dysplasia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86881

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Buschke-Ollendorff syndrome in a three-generation family: influence of a novel LEMD3 mutation to tropoelastin expression.
Burger B
European Journal of Dermatology, 20, 693-697 (2010)
Nuclear envelope protein MAN1 regulates clock through BMAL1.
Lin ST
eLife, 3, e02981-e02981 (2014)
Novel Somatic Mutation in LEMD3 Splice Site Results in Buschke-Ollendorff Syndrome with Polyostotic Melorheostosis and Osteopoikilosis.
Gutierrez D
Pediatric Dermatology, 32, e219-e220 (2015)
Marina Vietri et al.
Nature cell biology, 22(7), 856-867 (2020-07-01)
The ESCRT-III membrane fission machinery maintains the integrity of the nuclear envelope. Although primary nuclei resealing takes minutes, micronuclear envelope ruptures seem to be irreversible. Instead, micronuclear ruptures result in catastrophic membrane collapse and are associated with chromosome fragmentation and
Inhibition of TGF-? signaling at the nuclear envelope: characterization of interactions between MAN1, Smad2 and Smad3, and PPM1A.
Bourgeois B
Science Signaling, 6, ra49-ra49 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service