Skip to Content
Merck
All Photos(2)

Key Documents

HPA024729

Sigma-Aldrich

Anti-SLC28A3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Concentrative Na(+, Anti-Solute carrier family 28 member 3, Anti-hCNT3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

INCHHVLENAFNSTFPGNTTKVIACCQSLLSSTVAKGPGEVIPGGNHSLYSLKGCCTLLNPSTFNCNGISNT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC28A3(64078)

General description

The gene SLC28A3 (solute carrier family 28 member 3) is mapped to human chromosome 9q22. It is expressed in the pancreas, bone marrow, trachea, mammary gland, liver, prostate, intestine, brain and heart. The protein has 13 putative transmembrane helices.

Immunogen

Solute carrier family 28 member 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SLC28A3 (solute carrier family 28 member 3) is a Na+-dependent purine and pyrimidine nucleoside symporter. It also transports nucleoside drugs and analogs.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75460

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachel A Hesler et al.
Carcinogenesis, 37(11), 1041-1051 (2016-09-09)
Pancreatic ductal adenocarcinoma (PDAC) is a lethal cancer in part due to inherent resistance to chemotherapy, including the first-line drug gemcitabine. Although low expression of the nucleoside transporters hENT1 and hCNT3 that mediate cellular uptake of gemcitabine has been linked
Takashi Yamamoto et al.
European journal of pharmacology, 557(1), 1-8 (2006-12-05)
We investigated the mechanism of the transport of ribavirin (1-beta-D-ribofuranosyl-1,2,4-trizole-3-carboxamide) into placental epithelial cells using human choriocarcinoma (BeWo) cells and Xenopus oocytes expressing human nucleoside transporters. In BeWo cells, when a relatively low concentration (123 nM) of ribavirin was used
M W Ritzel et al.
The Journal of biological chemistry, 276(4), 2914-2927 (2000-10-18)
The human concentrative (Na(+)-linked) plasma membrane transport proteins hCNT1 and hCNT2 are selective for pyrimidine nucleosides (system cit) and purine nucleosides (system cif), respectively. Both have homologs in other mammalian species and belong to a gene family (CNT) that also
Huankai Hu et al.
Molecular pharmacology, 69(5), 1542-1553 (2006-02-01)
We characterized the electrophysiology, kinetics, and quantitative structure-activity relationship (QSAR) of the human concentrative nucleoside transporter 3 (hCNT3) expressed in Xenopus laevis oocytes by measuring substrate-induced inward currents using a two-microelectrode voltage-clamp system. At membrane potentials between -30 and -150

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service