Skip to Content
Merck
All Photos(2)

Key Documents

HPA024632

Sigma-Aldrich

Anti-TARBP1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Probable methyltransferase TARBP1, Anti-TAR RNA-binding protein 1, Anti-TAR RNA-binding protein of 185 kDa, Anti-TRP-185

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

MNELNSVSDLDRCHLYLMVLTELINLHLKVGWKRGNPIWRVISLLKNASIQHLQEMDSGQEPTVGSQIQRVVSMAALAMVCEAIDQKPELQLDSLHAG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TARBP1(6894)

General description

The gene TARBP1 (TAR (HIV-1) RNA binding protein 1) is mapped to human chromosome 1q42.2. The protein has dsRBDs (double stranded RNA binding domains), a KR motif in the dsRBD2 and a Medipal domain.

Immunogen

Probable methyltransferase TARBP1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TARBP1 (TAR (HIV-1) RNA binding protein 1) binds strongly to the small hairpin trans-activation responsive (TAR) RNA of HIV (human immunodeficiency virus)-1. It might have a role in disengaging RNA polymerase II from HIV-1 TAR RNA. In addition, it is also a part of the RNA-induced silencing complex (RISC). It interacts with Dicer and helps in processing of pre-microRNAs. TARBP1 also participates in other cellular functions, such as protein synthesis, RNA post-translational modifications, cellular proliferation, gene expression and stress-associated cellular processes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73865

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabel Freund et al.
RNA (New York, N.Y.), 25(7), 869-880 (2019-04-26)
Bacterial RNA has emerged as an important activator of innate immune responses by stimulating Toll-like receptors TLR7 and TLR8 in humans. Guanosine 2'-O-methylation at position 18 (Gm18) in bacterial tRNA was shown to antagonize tRNA-induced TLR7/8 activation, suggesting a potential
Robert D Beech et al.
Alcoholism, clinical and experimental research, 38(11), 2743-2753 (2014-11-26)
Alterations in stress-related gene expression may play a role in stress-related drinking and the risk of alcohol dependence. Microarrays were used to measure changes in gene expression in peripheral blood in nonsmoking, social drinking subjects exposed to 3 types of
Orla M Keane et al.
BMC genomics, 7, 42-42 (2006-03-07)
Gastrointestinal nematodes constitute a major cause of morbidity and mortality in grazing ruminants. Individual animals or breeds, however, are known to differ in their resistance to infection. Gene expression profiling allows us to examine large numbers of transcripts simultaneously in
F Wu-Baer et al.
The EMBO journal, 14(23), 5995-6009 (1995-12-01)
Activation of HIV-1 gene expression by the transactivator Tat is dependent on an RNA regulatory element located downstream of the transcription initiation site known as TAR. To characterize cellular factors that bind to TAR RNA and are involved in the
Sylvanne M Daniels et al.
BMC molecular biology, 10, 38-38 (2009-05-09)
Dicer, Ago2 and TRBP are the minimum components of the human RNA-induced silencing complex (RISC). While Dicer and Ago2 are RNases, TRBP is the double-stranded RNA binding protein (dsRBP) that loads small interfering RNA into the RISC. TRBP binds directly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service