Skip to Content
Merck
All Photos(4)

Key Documents

HPA023235

Sigma-Aldrich

Anti-CPQ antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-LDP, Anti-PGCP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

YERLALLVDTVGPRLSGSKNLEKAIQIMYQNLQQDGLEKVHLEPVRIPHWERGEESAVMLEPRIHKIAILGLGSSIGTPPE

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CPQ(10404)

General description

CPQ (carboxypeptidase Q) is mainly present in the placenta, kidney and blood plasma. It is a glutamate carboxypeptidase. The gene is mapped to human chromosome 8q21.

Immunogen

Plasma glutamate carboxypeptidase Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CPQ (carboxypeptidase Q) might be involved in the hydrolysis of circulating peptides. It participates in the breakdown of N-acetyl-l-aspartyl-l-glutamate into N-acetyl aspartate (NAA) and glutamate, thereby regulating glutamate homeostasis. It is upregulated in Hepatitis C virus-mediated hepatocellular carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76209

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jae-Hye Lee et al.
Oncotarget, 7(48), 79774-79786 (2016-11-03)
Tumor metastasis is the leading cause of cancer death. In the metastatic process, EMT is a unique phenotypic change that plays an important role in cell invasion and changes in cell morphology. Despite the clinical significance, the mechanism underlying tumor
F Michael Cutrer et al.
Headache, 53(2), 401-412 (2013-01-03)
The expansion of technologies available for the study of migraine pathophysiology has evolved greatly over the last 15 years. Two areas of rapid progress are investigations focusing on the genetics of migraine and others utilizing novel functional neuroimaging techniques. Genetic
Evan K Noch et al.
Advances in cancer research, 120, 159-192 (2013-07-31)
Astrocyte-elevated gene-1 (AEG-1/MTDH/LYRIC) is a potent oncogene that regulates key cellular processes underlying disease of the central nervous system (CNS). From its involvement in human immunodeficiency virus (HIV)-1 infection to its role in neurodegenerative disease and malignant brain tumors, AEG-1/MTDH/LYRIC
Maria W Smith et al.
Cancer research, 63(4), 859-864 (2003-02-20)
Hepatocellular carcinoma (HCC) is a common primary cancer associated frequently with hepatitis C virus (HCV). To gain insight into the molecular mechanisms of hepatocarcinogenesis, and to identify potential HCC markers, we performed cDNA microarray analysis on surgical liver samples from
R Gingras et al.
The Journal of biological chemistry, 274(17), 11742-11750 (1999-04-17)
We describe the identification, cDNA cloning, and biochemical characterization of a new human blood plasma glutamate carboxypeptidase (PGCP). PGCP was co-purified from human placenta with lysosomal carboxypeptidase, cathepsin A, lysosomal endopeptidase, cathepsin D, and a gamma-interferon-inducible protein, IP-30, using an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service