Skip to Content
Merck
All Photos(10)

Key Documents

HPA017330

Sigma-Aldrich

Anti-CALD1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CDM antibody produced in rabbit, Anti-Caldesmon antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

MTHKLKHTENTFSRPGGRASVDTKEAEGAPQVEAGKRLEELRRRRGETESEEFEKLKQKQQEAALELEELKKKREERRKVLEEEEQRRKQEEADRKLREEEEKRRLKEEIERRRAEAAEKRQKMPEDGLSDD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CALD1(800)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Caldesmon recombinant protein epitope signature tag (PrEST)

Application

Anti-CALD1 antibody produced in rabbit has been used in sandwich immunoassay of plasma samples.
Anti-CALD1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71213

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ulrika Qundos et al.
Journal of proteomics, 95, 46-54 (2013-05-02)
Several biobanking initiatives have emerged to create extensive collections of specimen for biomedical studies and various analytical platforms. An affinity proteomic analysis with antibody suspension bead arrays was conducted to investigate the influence of the pre-analytical time and temperature conditions
Christoph Köhler
Virchows Archiv : an international journal of pathology, 459(1), 81-89 (2011-06-01)
Alternative splicing of the caldesmon gene results in high (h-caldesmon) and low (l-caldesmon) molecular weight isoforms of the cytoskeleton-associated protein caldesmon. h-Caldesmon is predominantly expressed not only in smooth-muscle cells but also in pericryptal fibroblasts in colon. l-Caldesmon is widely
Christoph N Köhler
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 59(5), 504-517 (2011-03-18)
The author has recently reported the distribution of the cytoskeleton-associated protein caldesmon in spleen and lymph nodes detected with different antibodies against caldesmon (J Histochem Cytochem 58:183-193, 2010). Here the author reports the distribution of caldesmon in the CNS and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service