Skip to Content
Merck
All Photos(6)

Key Documents

HPA012867

Sigma-Aldrich

Anti-HTR2B antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-5-HT-2B, Anti-5-hydroxytryptamine receptor 2B, Anti-Serotonin receptor 2B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

TIHALQKKAYLVKNKPPQRLTWLTVSTVFQRDETPCSSPEKVAMLDGSRKDKALPNSGDETLMRRTSTIGKKSVQTISNEQRASKVL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HTR2B(3357)

General description

5-hydroxytryptamine receptor 2B (HTR2B) is a 481 amino acid receptor which is expressed in the liver, kidney, lungs and heart. The gene encoding this protein in located on chromosome 2q36.3-2q37.1.

Immunogen

5-hydroxytryptamine receptor 2B recombinant protein epitope signature tag (PrEST)

Application

Anti-HTR2B antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

HTR2B (5-hydroxytryptamine receptor 2B) is G-protein coupled receptor for 5-hydroxytryptamine (serotonin). An increased expression of this receptor is seen in idiopathic pulmonary fibrosis. The activated HTR2B regulates intracellular calcium levels and affects the activity of phosphatidylinositol 3-kinase. The pathway also involved in the activation of ryanodine receptors. It has been shown that HTR2B takes part in brain development and contributes to the brain architecture.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72721

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaëtan Le-Bel et al.
Experimental eye research, 184, 72-77 (2019-04-20)
Uveal melanoma (UM), although a very rare disease, remains a particularly aggressive type of cancer as near 50% of the UM presenting patients will also develop liver metastases within 15 years from the initial diagnostic. One of the most reliable
T T Vellinga et al.
Oncogene, 35(40), 5263-5271 (2016-03-22)
Gene expression-based classification systems have identified an aggressive colon cancer subtype with mesenchymal features, possibly reflecting epithelial-to-mesenchymal transition (EMT) of tumor cells. However, stromal fibroblasts contribute extensively to the mesenchymal phenotype of aggressive colon tumors, challenging the notion of tumor
Zhicheng Lin et al.
Pharmacogenetics, 14(12), 805-811 (2004-12-21)
5-Hydroxytryptamine (serotonin) receptor 2B (HTR2B) is involved in brain development. Although expressed in the human brain, HTR2B has not been investigated much for its role in higher brain functions. Here we describe a genome-scan with 391 simple sequence repeat markers
Ji Won Park et al.
British journal of cancer, 123(4), 610-618 (2020-06-04)
Immunomodulatory cytokines and systemic inflammatory markers are important during cancer development and progression. This study investigated the association and prognostic impact of systemic cytokine profiles and inflammatory markers in colorectal cancer (CRC). Interleukin (IL)-1β, IL-6, IL-8, IL-9, IL-10, tumour necrosis
Rohit N Kulkarni et al.
Diabetes, 61(9), 2205-2213 (2012-07-04)
A major goal in diabetes research is to find ways to enhance the mass and function of insulin secreting β-cells in the endocrine pancreas to prevent and/or delay the onset or even reverse overt diabetes. In this Perspectives in Diabetes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service