Skip to Content
Merck
All Photos(6)

Documents

HPA009067

Sigma-Aldrich

Anti-SPINK5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-LEKTI antibody produced in rabbit, Anti-Lympho-epithelial Kazal-type-related inhibitor antibody produced in rabbit, Anti-Serine protease inhibitor Kazal-type 5 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

DGRLGCTRENDPVLGPDGKTHGNKCAMCAELFLKEAENAKREGETRIRRNAEKDFCKEYEKQVRNGRLFCTRESDPVRGPDGRMHGNKCALCAEIFKQRFSEENSKTDQN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SPINK5(11005)

Looking for similar products? Visit Product Comparison Guide

General description

SPINK5 (serine peptidase inhibitor, Kazal type 5) gene is localized to human chromosome 5q32, which spans 61kb and contains 33exons. The encoded protein is expressed in oral mucosa, tonsils and thymus. It is also known as lymphoepithelial Kazal-type related inhibitor (LEKTI) and is a multi-domain protein.

Immunogen

Serine protease inhibitor Kazal-type 5 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SPINK5 (serine peptidase inhibitor, Kazal type 5) is involved in epithila formation where it controls proteolysis. It also plays an essential role in the terminal differentiation of keratinocytes. Mutations in this gene are linked with abnormalities in skin barrier. This gene is also associated with atopic diseases such as atopic dermatitis and asthma. Loss-of-function mutations in this gene result in autosomal recessive ichthyosiform disorder called Netherton syndrome, which is characterized by congenital scaly erytroderma, severe itching, eczema-like rashes, and recurrent asthma and food allergies. Down-regulation of this protein is linked with chronic rhinosinusitis, and is found in patients with intolerance to aspirin.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70809

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Márcia Gaião Alves et al.
Translational oncology, 14(1), 100970-100970 (2020-12-02)
Oral squamous cell carcinoma (OSCC) remains a challenging cancer to treat despite all the advances of the last 50 years. Kallikrein 5 (KLK5) is among the serine proteases implicated in OSCC development. However, whether the activity of KLK5 promotes carcinogenesis
Elina Eränkö et al.
Orphanet journal of rare diseases, 13(1), 213-213 (2018-11-28)
Netherton syndrome (NS) is a rare life-threatening syndrome caused by SPINK5 mutations leading to a skin barrier defect and a severe atopic diathesis. NS patients are prone to bacterial infections, but the understanding of the underlying immune deficiency is incomplete.
Elaine Zayas Marcelino da Silva et al.
Cancers, 13(17) (2021-09-11)
Head and neck squamous cell carcinoma remains challenging to treat with no improvement in survival rates over the past 50 years. Thus, there is an urgent need to discover more reliable therapeutic targets and biomarkers for HNSCC. Matriptase, a type-II
Y Nishio et al.
Genes and immunity, 4(7), 515-517 (2003-10-11)
Atopy, which is characterized by increased levels of immunoglobulin E (IgE) against common environmental allergens, is considered the strongest predisposing factor for asthma and atopic dermatitis (AD). Mutations in the gene encoding serine protease inhibitor Kazal-type 5 (SPINK5) are responsible
Kai Fruth et al.
The Laryngoscope, 122(6), 1198-1204 (2012-05-10)
Chronic rhinosinusitis (CRS) is a multifactorial disease that probably arises as a result of genetic diversity and environmental factors. SPINK5 is a serine protease inhibitor, which is supposed to be an important regulator of epithelial barrier maintenance. The role of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service