Skip to Content
Merck
All Photos(7)

Documents

HPA007266

Sigma-Aldrich

Anti-CAMK1D antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CKLiK antibody produced in rabbit, Anti-CaM kinase I δ antibody produced in rabbit, Anti-CaM kinase ID antibody produced in rabbit, Anti-CaM-KI δ antibody produced in rabbit, Anti-CaMKI δ antibody produced in rabbit, Anti-CaMKI-δ antibody produced in rabbit, Anti-Calcium/calmodulin-dependent protein kinase type 1D antibody produced in rabbit, Anti-CamKI-like protein kinase antibody produced in rabbit, Anti-Camk1D antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

MARENGESSSSWKKQAEDIKKIFEFKETLGTGAFSEVVLAEEKATGKLFAVKCIPKKALKGKESSIENEIAVLRKIKHENIVALEDIYESPNHLYLVMQLVSGG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAMK1D(57118)

Immunogen

Calcium/calmodulin-dependent protein kinase type 1D recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CAMK1D (Calcium/calmodulin-dependent protein kinase ID) is a novel granulocyte Ca++/calmodulin-dependent kinase responsible for controlling neuronal activities. It has four isoforms (α, β, γ, and δ). It drives the intracellular Ca2+ influxes via translocation in response to extracellular stimuli. It exerts Ca++ and calmodulin dependent kinase activity during the regulation of human granulocyte functions. It has been shown that during neutrophilic differentiation of CD34+ stem cells, the protein expression is upregulated in cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70186

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Verploegen et al.
Blood, 96(9), 3215-3223 (2000-10-26)
Human granulocytes are characterized by a variety of specific effector functions involved in host defense. Several widely expressed protein kinases have been implicated in the regulation of these effector functions. A polymerase chain reaction-based strategy was used to identify novel
Hiroyuki Sakagami et al.
The European journal of neuroscience, 22(11), 2697-2707 (2005-12-06)
Multifunctional Ca2+/calmodulin-dependent protein kinases (CaMKs) including CaMKI, II and IV, are thought to regulate a variety of neuronal functions. Unlike CaMKII, which is regulated by autophosphorylation, CaMKI as well as CaMKIV are activated by CaMKK. In this study, we examined
Akifumi Kamata et al.
Neuroscience research, 57(1), 86-97 (2006-10-24)
Among multifunctional Ca(2+)/calmodulin-dependent protein kinases (CaMKs), CaMKI has been shown to comprise a family of four structurally related isoforms (alpha, beta, gamma, and delta) encoded by separate genes with abundant expression in mature brain. In this study, we first examined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service