Skip to Content
Merck
All Photos(7)

Key Documents

HPA006885

Sigma-Aldrich

Anti-VIL1 antibody produced in rabbit

enhanced validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Villin-1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500
western blot: 0.04-0.4 μg/mL

immunogen sequence

NGPESTRMERLRGMTLAKEIRDQERGGRTYVGVVDGENELASPKLMEVMNHVLGKRRELKAAVPDTVVEPALKAALKLYHVSDSEGNLVVREVATRPLTQDLLSHE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VIL1(7429)

Looking for similar products? Visit Product Comparison Guide

General description

VIL1 (villin 1) is a gastrointestinal-related cyto-skeletal protein attached with the actin core bundle of brush border microvilli. It is expressed in the various epithelial cell lines such as intestinal mucosa, gall bladder, renal tubule cells, ductuli efferentes of the testis.

Immunogen

Villin-1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

VIL1 (villin 1) is associated with several cellular processes including actin cytoskeleton assembly, cell morphology, anti-apoptotic activities, and epithelial-to-mesenchymal transition. It is the main building block of cytoskeleton. In the presence of calcium, VIL1 binds to the microfilaments of actin. It acts as a potential diagnostic marker for several cancers such as cervical adenocarcinoma, endometrial adenocarcinoma and α-fetoprotein-associated hepatocellular carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70146

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nick M Shillingford et al.
The American journal of surgical pathology, 39(2), 245-250 (2014-12-18)
Microvillus inclusion disease (MVID) is a rare congenital disorder that manifests early in infancy as intractable watery diarrhea. The entity is characterized morphologically by a deficient brush border and apical cytoplasmic inclusions within absorptive cells (enterocytes) due to misplaced assembly
Etsuko Nakamura et al.
Cancer biology & therapy, 12(3), 181-190 (2011-05-31)
Villin1 (VIL1) has a role in regulating actin dynamics, cell morphology, anti-apoptotic mechanisms, and epithelial-to-mesenchymal transition. Previously we reported VIL1 as a novel diagnostic marker for cervical adenocarcinoma (AC) with poor radioresponse. This study further investigated the diagnostic role of
Maimaiti Xieraili et al.
Cancer science, 103(8), 1493-1501 (2012-04-26)
The prognostic assessment of patients with hepatocellular carcinoma (HCC) after resection is an important clinical issue. The present study investigated those genes associated with high serum alpha-fetoprotein (AFP), and their clinical significance, including prognosis and recurrence after hepatectomy. Based on
Etsuko Nakamura et al.
Cancer biology & therapy, 8(12), 1146-1153 (2009-04-21)
The number of new cervical adenocarcinoma (AD) cases has risen slowly, however, its histological similarity to other tumor types and the difficulty of identifying the site of the original tumor makes the diagnosis of cervical AD particularly challenging. We investigated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service