Skip to Content
Merck
All Photos(1)

Key Documents

H2380

Sigma-Aldrich

6-Hydroxy-DL-DOPA

≥98% (HPLC), powder, APE1 inhibitor

Synonym(s):

2,4,5-Trihydroxy-DL-phenylalanine, 2,5-Dihydroxy-DL-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO5
CAS Number:
Molecular Weight:
213.19
Beilstein:
2860065
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

product name

6-Hydroxy-DL-DOPA, ≥98% (HPLC), powder

Assay

≥98% (HPLC)

form

powder

color

off-white

solubility

H2O: 3 mg/mL
1 M HCl: 50 mg/mL (Solutions should be freshly prepared and protected from exposure to light.)

storage temp.

−20°C

SMILES string

NC(Cc1cc(O)c(O)cc1O)C(O)=O

InChI

1S/C9H11NO5/c10-5(9(14)15)1-4-2-7(12)8(13)3-6(4)11/h2-3,5,11-13H,1,10H2,(H,14,15)

InChI key

YLKRUSPZOTYMAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dissolve in oxygen-free boiled water containing 0.1% sodium metabisulfite or other antioxidant. Solutions should be freshly prepared and protected from exposure to light.

Biochem/physiol Actions

6-Hydroxy-DL-DOPA is an inhibitor of APE1 (apurinic/apyrimidinic endonuclease) repair-function activity. APE1 is a nuclease crucial for DNA base excision repair pathway. 6-Hydroxy-DL-DOPA also blocks RAD52 (DNA repair protein) single-stranded DNA binding domain.
Precursor of the catecholaminergic neurotoxin, 6-hydroxydopamine; converted to 6-hydroxydopamine by L-aromatic amino acid decarboxylase.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Caution

Hygroscopic; photosensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gurushankar Chandramouly et al.
Chemistry & biology, 22(11), 1491-1504 (2015-11-10)
Suppression of RAD52 causes synthetic lethality in BRCA-deficient cells. Yet pharmacological inhibition of RAD52, which binds single-strand DNA (ssDNA) and lacks enzymatic activity, has not been demonstrated. Here, we identify the small molecule 6-hydroxy-DL-dopa (6-OH-dopa) as a major allosteric inhibitor
Adnan Azeem et al.
International journal of pharmaceutics, 422(1-2), 436-444 (2011-11-08)
Ropinirole, a recent introduction in the clinical treatment of Parkinson's disease, suffers with the problems of low oral bioavailability and frequent dosing. An effective transdermal nano-emulsion drug delivery system can however resolve these issues effectively with greater therapeutic benefits and
A J Nappi et al.
Biochimica et biophysica acta, 1201(3), 498-504 (1994-12-15)
The interactions of ascorbic acid (AA) and reduced glutathione (GSH) in the oxidations of the catecholaminergic neurotoxins 6-hydroxydopa (TOPA) and 6-hydroxydopamine (6-OHDA) were investigated by both high performance liquid chromatography with electrochemical detection (HPLC-ED) and spectrometric methods. These comparative studies
Annalisa Pinna et al.
Synapse (New York, N.Y.), 61(8), 606-614 (2007-05-04)
Evidence obtained in rodent and primate models of Parkinson's disease (PD) and preliminary clinical trials, indicates that adenosine A(2A) receptor antagonists might represent a promising nondopaminergic therapeutic tool for the treatment of PD. Those studies demonstrated the ability of adenosine
R Medda et al.
Biochemistry, 34(50), 16375-16381 (1995-12-19)
Intermediate states in the catalytic mechanism of lentil copper amine oxidase have been investigated by ESR and optical spectroscopy. Using highly purified apo- and holoenzyme in combination with a poor substrate and a range of inhibitors, under both aerobic and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service