Skip to Content
Merck
All Photos(1)

Key Documents

H0377

Sigma-Aldrich

DL-5-Hydroxylysine hydrochloride

≥98% (TLC)

Synonym(s):

2,6-Diamino-5-hydroxycaproic acid hydrochloride, 2,6-Diamino-5-hydroxyhexanoic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH(OH)CH2CH2CH(NH2)COOH · HCl
CAS Number:
Molecular Weight:
198.65
Beilstein:
3914368
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

DL-5-Hydroxylysine hydrochloride,

Assay

≥98% (TLC)

Quality Level

form

powder

color

white

mp

225 °C (dec.) (lit.)

application(s)

detection
peptide synthesis

SMILES string

Cl.NCC(O)CCC(N)C(O)=O

InChI

1S/C6H14N2O3.ClH/c7-3-4(9)1-2-5(8)6(10)11;/h4-5,9H,1-3,7-8H2,(H,10,11);1H

InChI key

MJXVOTKVFFAZQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Characterization of acetyl-CoA: L-lysine N6-acetyltransferase, which catalyses the first step of carbon catabolism from lysine in Saccharomyces cerevisiae.: This research investigates the enzyme acetyl-CoA: L-lysine N6-acetyltransferase, which initiates the catabolism of lysine in Saccharomyces cerevisiae. Utilizing DL-5-Hydroxylysine hydrochloride, the study provides insights into the metabolic pathways and regulatory mechanisms of lysine degradation, contributing to the broader understanding of amino acid metabolism in yeast (Bode et al., 1993).

Biochem/physiol Actions

DL-5-Hydroxylysine is a racemic mixture of D- and L- enantiomers of 5-hydroxylysine which may be used as potential target markers for radical-induced protein oxidation.

Other Notes

Mixed DL and DL-allo

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E R Stadtman et al.
Amino acids, 25(3-4), 207-218 (2003-12-09)
We summarize here results of studies designed to elucidate basic mechanisms of reactive oxygen (ROS)-mediated oxidation of proteins and free amino acids. These studies have shown that oxidation of proteins can lead to hydroxylation of aromatic groups and aliphatic amino
R Bode et al.
Archives of microbiology, 160(5), 397-400 (1993-01-01)
The carbon catabolism of L-lysine starts in Saccharomyces cerevisiae with acetylation by an acetyl-CoA:L-lysine N6-acetyltransferase. The enzyme is strongly induced in cells grown on L-lysine as sole carbon source and has been purified about 530-fold. Its activity was specific for
B Morin et al.
Chemical research in toxicology, 11(11), 1265-1273 (1998-11-17)
gamma-Irradiation of several amino acids (Val, Leu, Ile, Lys, Pro, and Glu) in the presence of O2 generates hydroperoxides. We have previously isolated and characterized valine and leucine hydroperoxides, and hydroxides, and have detected these products in both isolated systems
A Rahman et al.
Plant & cell physiology, 41(1), 1-9 (2000-04-06)
Chromosaponin I (CSI), a triterpenoid saponin isolated from pea, stimulates the growth of roots in Arabidopsis thaliana seedlings on wetted filter paper in the light for 14 d. The growth rates of roots in Columbia (Col) and Landsberg erecta (Ler)
Giuseppe Forlani et al.
Frontiers in plant science, 11, 582026-582026 (2020-11-17)
Because proline accumulates rapidly in response to several stress conditions such as drought and excess salt, increased intracellular levels of free proline are considered a hallmark of adaptive reactions in plants, particularly in response to water stress. Proline quantitation is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service