Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5110-01

HiTrap® IGM purification

Cytiva 17-5110-01, pack of 5 × 1 mL

Synonym(s):

IGM Purification Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

ligand

2-mercaptopyridine

packaging

pack of 5 × 1 mL

manufacturer/tradename

Cytiva 17-5110-01

storage condition

(20% Ethanol)

parameter

<4 mL/min flow rate
42 psi

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

16 mm

matrix

highly cross-linked 6% agarose

avg. part. size

34 μm

cleaning

2-13

working range

3-11

storage temp.

2-8°C

General description

HiTrap® IgM Purification HP is a 1 mL column prepacked with Sepharose High Performancefor fast and efficient purification of monoclonal IgM.

Application

HiTrap® IgM Purification HP is prepacked with a thiophilic adsorption medium, 2-merCaptopyridine coupled to Sepharose High Performance. Thiophilic adsorption is promoted by water-structuring salts. It has been suggested that the interaction of IgM and ligand results from combined electron donating and accepting action of the ligand or, alternatively, a mixed mode hydrophilic-hydrophobic interaction.

Features and Benefits

  • Optimized for fast, efficient purification of monoclonal IgM from hybridoma cell culture.
  • Prepacked with a thiophilic adsorption medium, 2-merCaptopyridine coupled to Sepharose High Performance.
  • Recovery of IgM is high with excellent activity retention.
  • The top and bottom frits are manufactured from porous polyethylene
  • Columns are delivered with a stopper on the inlet and a snap-off end on the outle

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

HiTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service