GE17-1275-01
SOURCE™ 30Q
Cytiva 17-1275-01, pack of 50 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
ligand
quaternary ammonium
description
Ion Exchanger Type (value)
packaging
pack of 50 mL
manufacturer/tradename
Cytiva 17-1275-01
matrix
polystyrene/divinylbenzene
particle size
30 μm
cleaning in place
1-14
working range
2-12
suitability
suitable for bioprocess medium (BioProcess medium. Process-scale quantities are available.)
suitable for bioprocess medium
General description
SOURCE™ 30Q is a polymeric, strong anion exchanger designed for intermediate purification and large-scale polishing.
Application
SOURCE™ 30Q is a synthetic high performance, preparative, chromatography medium, based on a 30 μm monosized, rigid polystyrene/divinyl benzene polymer matrix. It is modified with quaternary ammonium (Q) strong anion exchange groups. SOURCE™ media have excellent physical and chemical characteristics, allowing high flow rates and consistent performance at both laboratory and process scales.
SOURCE™ 30Q is suitable for intermediate purification steps in industrial processes when large volumes of partially purified material need to be processed. A high degree of purification can be obtained with high productivity.
Through the combination of high quality assurance standards and a patented manufacturing process, particle structure is consistent both bead-to-bead and batch-to batch. As member of the BioProcess media range, SOURCE™ 30Q meets industrial demands with security of supply and comprehensive technical and regulatory support
SOURCE™ 30Q is suitable for intermediate purification steps in industrial processes when large volumes of partially purified material need to be processed. A high degree of purification can be obtained with high productivity.
Through the combination of high quality assurance standards and a patented manufacturing process, particle structure is consistent both bead-to-bead and batch-to batch. As member of the BioProcess media range, SOURCE™ 30Q meets industrial demands with security of supply and comprehensive technical and regulatory support
Features and Benefits
- Mono-sized 30 um rigid beads gives Low back pressure and high flow rates allowing for high productivity
- Strong anion exchanger designed for high resolution polishing purification of Proteins, peptides and oligonucleotides
- High chemical stability allowing for wide range of working conditions with good resistance to cleaning conditions at high pH
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
SOURCE is a trademark of Cytiva
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service